Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 64177214-64177483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROR1
Entrez Gene ID
4919
Gene Name
receptor tyrosine kinase like orphan receptor 1
Gene Aliases
NTRKR1, dJ537F10.1
UniGene
Hs.128753
Gene Chromosome Location
Chr.1: 63774019-64181498 on Build GRCh38
Transcripts
NM_005012.3 XM_011541526.1 XM_017001376.1 XM_017001377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
228795 c.1429G>A p.E477K Chr.1: 64177470-64177470 on Build GRCh38
911500 c.1420C>T p.R474C Chr.1: 64177461-64177461 on Build GRCh38
5907163 c.1399G>A p.E467K Chr.1: 64177440-64177440 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1001072903 Chr.1: 64177235-64177235 on Build GRCh38
rs540448964 Chr.1: 64177404-64177404 on Build GRCh38
rs150902668 Chr.1: 64177257-64177257 on Build GRCh38
rs1015502907 Chr.1: 64177353-64177353 on Build GRCh38
rs1030707863 Chr.1: 64177296-64177296 on Build GRCh38
rs752027399 Chr.1: 64177405-64177405 on Build GRCh38
rs950806203 Chr.1: 64177367-64177367 on Build GRCh38
rs368318432 Chr.1: 64177390-64177390 on Build GRCh38
rs746658433 Chr.1: 64177294-64177294 on Build GRCh38
rs760194372 Chr.1: 64177382-64177382 on Build GRCh38
rs996199446 Chr.1: 64177283-64177283 on Build GRCh38
rs750686747 Chr.1: 64177413-64177414 on Build GRCh38
rs754615045 Chr.1: 64177342-64177342 on Build GRCh38
rs961239707 Chr.1: 64177389-64177389 on Build GRCh38
rs367937524 Chr.1: 64177272-64177272 on Build GRCh38
rs919683295 Chr.1: 64177398-64177398 on Build GRCh38
rs765571751 Chr.1: 64177399-64177399 on Build GRCh38
rs573051336 Chr.1: 64177276-64177276 on Build GRCh38
rs780600688 Chr.1: 64177401-64177401 on Build GRCh38
rs781140056 Chr.1: 64177446-64177446 on Build GRCh38
rs1030554429 Chr.1: 64177364-64177364 on Build GRCh38
rs372007035 Chr.1: 64177403-64177403 on Build GRCh38
rs757647552 Chr.1: 64177352-64177352 on Build GRCh38
rs757301104 Chr.1: 64177445-64177445 on Build GRCh38
rs955335963 Chr.1: 64177321-64177321 on Build GRCh38
rs987092755 Chr.1: 64177325-64177325 on Build GRCh38
rs973893776 Chr.1: 64177392-64177392 on Build GRCh38
rs776279520 Chr.1: 64177393-64177393 on Build GRCh38
rs980124313 Chr.1: 64177406-64177406 on Build GRCh38
rs910864408 Chr.1: 64177237-64177237 on Build GRCh38
rs900584228 Chr.1: 64177244-64177244 on Build GRCh38

View Full Product Details