Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51332502-51332757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VSIG10L
Entrez Gene ID
147645
Gene Name
V-set and immunoglobulin domain containing 10 like
Gene Aliases
-
UniGene
Hs.293236
Gene Chromosome Location
Chr.19: 51331541-51342124 on Build GRCh38
Transcripts
NM_001163922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4285948 c.2575G>C p.A859P Chr.19: 51332640-51332640 on Build GRCh38
5891137 c.2582C>T p.T861I Chr.19: 51332633-51332633 on Build GRCh38
5022192 c.2576_2577insC p.Q860fs*>9 Chr.19: 51332638-51332639 on Build GRCh38
4602882 c.2577_2578insC p.Q860fs*>9 Chr.19: 51332637-51332638 on Build GRCh38
5446540 c.2575_2576insC p.Q860fs*>9 Chr.19: 51332639-51332640 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs543877327 Chr.19: 51332588-51332588 on Build GRCh38
rs201835166 Chr.19: 51332658-51332658 on Build GRCh38
rs200336358 Chr.19: 51332640-51332640 on Build GRCh38
rs1055075797 Chr.19: 51332557-51332557 on Build GRCh38
rs1046109563 Chr.19: 51332532-51332532 on Build GRCh38
rs899066595 Chr.19: 51332561-51332561 on Build GRCh38
rs758733818 Chr.19: 51332544-51332544 on Build GRCh38
rs886241263 Chr.19: 51332593-51332593 on Build GRCh38
rs541389662 Chr.19: 51332634-51332634 on Build GRCh38
rs372076222 Chr.19: 51332653-51332653 on Build GRCh38
rs751611081 Chr.19: 51332606-51332607 on Build GRCh38
rs780371154 Chr.19: 51332576-51332576 on Build GRCh38
rs397723022 Chr.19: 51332639-51332640 on Build GRCh38
rs748232414 Chr.19: 51332631-51332631 on Build GRCh38
rs28740866 Chr.19: 51332587-51332587 on Build GRCh38
rs373144536 Chr.19: 51332695-51332695 on Build GRCh38
rs916539684 Chr.19: 51332644-51332644 on Build GRCh38
rs927638405 Chr.19: 51332534-51332534 on Build GRCh38
rs903561417 Chr.19: 51332705-51332705 on Build GRCh38
rs11402251 Chr.19: 51332638-51332639 on Build GRCh38
rs370588522 Chr.19: 51332613-51332618 on Build GRCh38
rs750774904 Chr.19: 51332608-51332608 on Build GRCh38
rs76256168 Chr.19: 51332589-51332589 on Build GRCh38
rs372427820 Chr.19: 51332567-51332567 on Build GRCh38
rs869139085 Chr.19: 51332640-51332641 on Build GRCh38
rs749086734 Chr.19: 51332654-51332654 on Build GRCh38
rs6509526 Chr.19: 51332540-51332540 on Build GRCh38
rs935108761 Chr.19: 51332718-51332718 on Build GRCh38
rs769763868 Chr.19: 51332645-51332645 on Build GRCh38
rs117141091 Chr.19: 51332696-51332696 on Build GRCh38
rs951500263 Chr.19: 51332592-51332592 on Build GRCh38
rs184769661 Chr.19: 51332590-51332590 on Build GRCh38

View Full Product Details