Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1296159-1296430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNPH
Entrez Gene ID
9751
Gene Name
syntaphilin
Gene Aliases
-
UniGene
Hs.323833
Gene Chromosome Location
Chr.20: 1266291-1309327 on Build GRCh38
Transcripts
NM_001318234.1 NM_014723.3 XM_005260889.3 XM_011529402.2 XM_011529403.2 XM_011529404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1024544 c.114T>C p.L38L Chr.20: 1296353-1296353 on Build GRCh38
1024543 c.70C>T p.R24C Chr.20: 1296309-1296309 on Build GRCh38
4289602 c.66A>C p.P22P Chr.20: 1296305-1296305 on Build GRCh38
1024542 c.66A>G p.P22P Chr.20: 1296305-1296305 on Build GRCh38
6786142 c.1544G>A p.G515D Chr.20: 1296375-1296375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759021292 Chr.20: 1296284-1296284 on Build GRCh38
rs145237357 Chr.20: 1296331-1296331 on Build GRCh38
rs780246617 Chr.20: 1296385-1296385 on Build GRCh38
rs773170529 Chr.20: 1296329-1296329 on Build GRCh38
rs756311541 Chr.20: 1296378-1296378 on Build GRCh38
rs757443788 Chr.20: 1296307-1296307 on Build GRCh38
rs535039485 Chr.20: 1296391-1296391 on Build GRCh38
rs751884627 Chr.20: 1296354-1296354 on Build GRCh38
rs929641942 Chr.20: 1296299-1296299 on Build GRCh38
rs764537703 Chr.20: 1296350-1296350 on Build GRCh38
rs1054296323 Chr.20: 1296254-1296254 on Build GRCh38
rs867347470 Chr.20: 1296260-1296260 on Build GRCh38
rs773043905 Chr.20: 1296244-1296244 on Build GRCh38
rs73073678 Chr.20: 1296338-1296338 on Build GRCh38
rs781163152 Chr.20: 1296201-1296201 on Build GRCh38
rs376275699 Chr.20: 1296330-1296330 on Build GRCh38
rs773622068 Chr.20: 1296210-1296210 on Build GRCh38
rs753193400 Chr.20: 1296295-1296295 on Build GRCh38
rs549972502 Chr.20: 1296261-1296261 on Build GRCh38
rs569907857 Chr.20: 1296309-1296309 on Build GRCh38
rs367658252 Chr.20: 1296232-1296232 on Build GRCh38
rs1046296635 Chr.20: 1296316-1296316 on Build GRCh38
rs372864352 Chr.20: 1296407-1296407 on Build GRCh38
rs528071855 Chr.20: 1296227-1296227 on Build GRCh38
rs779050727 Chr.20: 1296319-1296319 on Build GRCh38
rs778080356 Chr.20: 1296304-1296304 on Build GRCh38
rs774362667 Chr.20: 1296219-1296219 on Build GRCh38
rs772028994 Chr.20: 1296327-1296327 on Build GRCh38
rs752196640 Chr.20: 1296238-1296238 on Build GRCh38
rs764708985 Chr.20: 1296339-1296339 on Build GRCh38
rs758812111 Chr.20: 1296303-1296303 on Build GRCh38
rs769912928 Chr.20: 1296317-1296317 on Build GRCh38
rs745913424 Chr.20: 1296208-1296208 on Build GRCh38
rs766012262 Chr.20: 1296252-1296252 on Build GRCh38
rs150438827 Chr.20: 1296245-1296245 on Build GRCh38
rs566005501 Chr.20: 1296358-1296358 on Build GRCh38
rs373921517 Chr.20: 1296310-1296310 on Build GRCh38
rs1010279134 Chr.20: 1296184-1296184 on Build GRCh38
rs758429519 Chr.20: 1296400-1296400 on Build GRCh38
rs367993627 Chr.20: 1296357-1296357 on Build GRCh38
rs776017391 Chr.20: 1296348-1296348 on Build GRCh38
rs765804483 Chr.20: 1296285-1296285 on Build GRCh38
rs374099562 Chr.20: 1296349-1296349 on Build GRCh38
rs776344518 Chr.20: 1296332-1296332 on Build GRCh38
rs957373087 Chr.20: 1296182-1296182 on Build GRCh38
rs748073256 Chr.20: 1296325-1296325 on Build GRCh38
rs751882983 Chr.20: 1296305-1296305 on Build GRCh38
rs564826578 Chr.20: 1296239-1296239 on Build GRCh38

View Full Product Details