Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 56891806-56892065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM25
Entrez Gene ID
7706
Gene Name
tripartite motif containing 25
Gene Aliases
EFP, RNF147, Z147, ZNF147
UniGene
Hs.528952
Gene Chromosome Location
Chr.17: 56887909-56914048 on Build GRCh38
Transcripts
NM_005082.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6807997 c.1557G>A p.Q519Q Chr.17: 56892036-56892036 on Build GRCh38
3983561 c.1679C>G p.A560G Chr.17: 56891914-56891914 on Build GRCh38
6556503 c.1727G>A p.R576Q Chr.17: 56891866-56891866 on Build GRCh38
7424763 c.1726C>T p.R576W Chr.17: 56891867-56891867 on Build GRCh38
6807999 c.1651G>A p.V551M Chr.17: 56891942-56891942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373822862 Chr.17: 56891964-56891964 on Build GRCh38
rs150493607 Chr.17: 56891955-56891955 on Build GRCh38
rs774609957 Chr.17: 56891861-56891861 on Build GRCh38
rs769094656 Chr.17: 56892030-56892030 on Build GRCh38
rs539288584 Chr.17: 56891991-56891991 on Build GRCh38
rs760096757 Chr.17: 56891866-56891866 on Build GRCh38
rs765845402 Chr.17: 56891882-56891882 on Build GRCh38
rs761145340 Chr.17: 56891956-56891956 on Build GRCh38
rs779281787 Chr.17: 56892009-56892009 on Build GRCh38
rs769870444 Chr.17: 56891838-56891838 on Build GRCh38
rs367715143 Chr.17: 56891989-56891989 on Build GRCh38
rs374799982 Chr.17: 56891828-56891828 on Build GRCh38
rs146675083 Chr.17: 56891900-56891900 on Build GRCh38
rs764516557 Chr.17: 56891959-56891959 on Build GRCh38
rs200806739 Chr.17: 56891926-56891926 on Build GRCh38
rs754492390 Chr.17: 56891973-56891973 on Build GRCh38
rs749116784 Chr.17: 56892002-56892002 on Build GRCh38
rs781223146 Chr.17: 56891885-56891885 on Build GRCh38
rs187826161 Chr.17: 56892003-56892003 on Build GRCh38
rs113318936 Chr.17: 56891943-56891943 on Build GRCh38
rs766991018 Chr.17: 56891982-56891982 on Build GRCh38
rs747521958 Chr.17: 56892024-56892024 on Build GRCh38
rs752961752 Chr.17: 56891867-56891867 on Build GRCh38
rs758113639 Chr.17: 56891879-56891879 on Build GRCh38
rs773493267 Chr.17: 56891847-56891847 on Build GRCh38
rs866223501 Chr.17: 56891966-56891966 on Build GRCh38
rs148351661 Chr.17: 56891901-56891901 on Build GRCh38
rs561236452 Chr.17: 56891904-56891904 on Build GRCh38
rs137888736 Chr.17: 56891967-56891967 on Build GRCh38
rs933287651 Chr.17: 56892027-56892027 on Build GRCh38
rs763140638 Chr.17: 56891848-56891848 on Build GRCh38
rs754865161 Chr.17: 56891883-56891883 on Build GRCh38
rs368145197 Chr.17: 56891929-56891929 on Build GRCh38
rs371770246 Chr.17: 56891962-56891962 on Build GRCh38
rs61738274 Chr.17: 56891868-56891868 on Build GRCh38
rs745885149 Chr.17: 56892018-56892018 on Build GRCh38
rs771125728 Chr.17: 56891850-56891850 on Build GRCh38
rs764739358 Chr.17: 56891869-56891869 on Build GRCh38
rs756042254 Chr.17: 56891990-56891990 on Build GRCh38
rs747833445 Chr.17: 56891886-56891886 on Build GRCh38
rs772179850 Chr.17: 56892020-56892020 on Build GRCh38
rs762576334 Chr.17: 56891963-56891963 on Build GRCh38
rs775499448 Chr.17: 56892022-56892022 on Build GRCh38
rs1023121562 Chr.17: 56891895-56891895 on Build GRCh38
rs755921273 Chr.17: 56891890-56891890 on Build GRCh38
rs750078996 Chr.17: 56891878-56891878 on Build GRCh38

View Full Product Details