Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58358526-58358799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF43
Entrez Gene ID
54894
Gene Name
ring finger protein 43
Gene Aliases
RNF124, URCC
UniGene
Hs.584916
Gene Chromosome Location
Chr.17: 58352500-58418895 on Build GRCh38
Transcripts
NM_001305544.1 NM_001305545.1 NM_017763.5 XM_011524955.2 XM_011524956.2 XM_017024800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3787382 c.1189C>T p.R397W Chr.17: 58358587-58358587 on Build GRCh38
4390103 c.1181C>T p.A394V Chr.17: 58358595-58358595 on Build GRCh38
4651313 c.1165C>T p.R389C Chr.17: 58358611-58358611 on Build GRCh38
6932604 c.1040G>A p.G347D Chr.17: 58358736-58358736 on Build GRCh38
6981612 c.1220G>C p.G407A Chr.17: 58358556-58358556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919729695 Chr.17: 58358625-58358625 on Build GRCh38
rs771831816 Chr.17: 58358665-58358665 on Build GRCh38
rs866759587 Chr.17: 58358651-58358651 on Build GRCh38
rs1011735047 Chr.17: 58358709-58358709 on Build GRCh38
rs367829905 Chr.17: 58358645-58358645 on Build GRCh38
rs747686258 Chr.17: 58358611-58358611 on Build GRCh38
rs778993511 Chr.17: 58358758-58358758 on Build GRCh38
rs745728704 Chr.17: 58358759-58358759 on Build GRCh38
rs564401086 Chr.17: 58358559-58358559 on Build GRCh38
rs199598395 Chr.17: 58358769-58358769 on Build GRCh38
rs929720505 Chr.17: 58358664-58358664 on Build GRCh38
rs568220750 Chr.17: 58358674-58358674 on Build GRCh38
rs760806622 Chr.17: 58358584-58358584 on Build GRCh38
rs887022726 Chr.17: 58358654-58358654 on Build GRCh38
rs34523089 Chr.17: 58358748-58358748 on Build GRCh38
rs755684321 Chr.17: 58358747-58358747 on Build GRCh38
rs549993660 Chr.17: 58358597-58358597 on Build GRCh38
rs769498996 Chr.17: 58358619-58358619 on Build GRCh38
rs770626824 Chr.17: 58358756-58358756 on Build GRCh38
rs780224400 Chr.17: 58358737-58358737 on Build GRCh38
rs138844366 Chr.17: 58358766-58358766 on Build GRCh38
rs774483341 Chr.17: 58358653-58358653 on Build GRCh38
rs779431648 Chr.17: 58358599-58358599 on Build GRCh38
rs531889929 Chr.17: 58358587-58358587 on Build GRCh38
rs777370175 Chr.17: 58358620-58358620 on Build GRCh38
rs775363695 Chr.17: 58358673-58358673 on Build GRCh38
rs767761157 Chr.17: 58358566-58358566 on Build GRCh38
rs990372544 Chr.17: 58358575-58358575 on Build GRCh38
rs758947339 Chr.17: 58358719-58358719 on Build GRCh38
rs750959307 Chr.17: 58358707-58358707 on Build GRCh38
rs1020581544 Chr.17: 58358726-58358726 on Build GRCh38
rs1029129424 Chr.17: 58358544-58358544 on Build GRCh38
rs758746256 Chr.17: 58358605-58358605 on Build GRCh38
rs376002987 Chr.17: 58358688-58358688 on Build GRCh38
rs947231021 Chr.17: 58358598-58358598 on Build GRCh38
rs369745984 Chr.17: 58358749-58358749 on Build GRCh38
rs79309393 Chr.17: 58358683-58358683 on Build GRCh38
rs750759880 Chr.17: 58358600-58358600 on Build GRCh38
rs759290047 Chr.17: 58358565-58358565 on Build GRCh38
rs766444227 Chr.17: 58358550-58358550 on Build GRCh38
rs978656819 Chr.17: 58358609-58358609 on Build GRCh38
rs751977513 Chr.17: 58358744-58358744 on Build GRCh38
rs201204271 Chr.17: 58358662-58358662 on Build GRCh38
rs752924746 Chr.17: 58358582-58358582 on Build GRCh38
rs780893183 Chr.17: 58358610-58358610 on Build GRCh38
rs981456911 Chr.17: 58358591-58358591 on Build GRCh38
rs748840036 Chr.17: 58358643-58358643 on Build GRCh38
rs145642181 Chr.17: 58358681-58358681 on Build GRCh38
rs761926947 Chr.17: 58358689-58358689 on Build GRCh38
rs764017391 Chr.17: 58358586-58358586 on Build GRCh38

View Full Product Details