Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75398587-75398826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF503
Entrez Gene ID
84858
Gene Name
zinc finger protein 503
Gene Aliases
NOLZ-1, NOLZ1, Nlz2
UniGene
Hs.195710
Gene Chromosome Location
Chr.10: 75279726-75401894 on Build GRCh38
Transcripts
NM_032772.5 NR_120651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1349189 c.1931G>T p.G644V Chr.10: 75398759-75398759 on Build GRCh38
80791 c.1872A>C p.G624G Chr.10: 75398818-75398818 on Build GRCh38
1603748 c.1900G>T p.G634* Chr.10: 75398790-75398790 on Build GRCh38
6518020 c.1864G>T p.A622S Chr.10: 75398826-75398826 on Build GRCh38
3440695 c.1874C>G p.P625R Chr.10: 75398816-75398816 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182331762 Chr.10: 75398651-75398651 on Build GRCh38
rs763565405 Chr.10: 75398798-75398798 on Build GRCh38
rs1014129585 Chr.10: 75398613-75398613 on Build GRCh38
rs749287676 Chr.10: 75398755-75398755 on Build GRCh38
rs368925089 Chr.10: 75398794-75398794 on Build GRCh38
rs755668965 Chr.10: 75398725-75398725 on Build GRCh38
rs933255185 Chr.10: 75398763-75398763 on Build GRCh38
rs535101391 Chr.10: 75398715-75398715 on Build GRCh38
rs1056388759 Chr.10: 75398728-75398728 on Build GRCh38
rs761143408 Chr.10: 75398800-75398808 on Build GRCh38
rs759178576 Chr.10: 75398804-75398804 on Build GRCh38
rs747188548 Chr.10: 75398779-75398779 on Build GRCh38
rs780709304 Chr.10: 75398714-75398714 on Build GRCh38
rs896445650 Chr.10: 75398739-75398739 on Build GRCh38
rs538172238 Chr.10: 75398619-75398619 on Build GRCh38
rs950855924 Chr.10: 75398650-75398650 on Build GRCh38
rs577000468 Chr.10: 75398800-75398800 on Build GRCh38
rs770701075 Chr.10: 75398765-75398765 on Build GRCh38
rs568088470 Chr.10: 75398683-75398683 on Build GRCh38
rs983565724 Chr.10: 75398655-75398655 on Build GRCh38
rs1016257359 Chr.10: 75398674-75398674 on Build GRCh38
rs979609190 Chr.10: 75398656-75398656 on Build GRCh38
rs776727778 Chr.10: 75398790-75398790 on Build GRCh38
rs770037190 Chr.10: 75398796-75398796 on Build GRCh38
rs1045042390 Chr.10: 75398745-75398745 on Build GRCh38
rs769830389 Chr.10: 75398710-75398710 on Build GRCh38
rs76853207 Chr.10: 75398676-75398676 on Build GRCh38
rs774605653 Chr.10: 75398803-75398803 on Build GRCh38
rs558746353 Chr.10: 75398797-75398797 on Build GRCh38
rs943418948 Chr.10: 75398680-75398680 on Build GRCh38
rs746127513 Chr.10: 75398773-75398773 on Build GRCh38
rs778587682 Chr.10: 75398776-75398776 on Build GRCh38
rs572054889 Chr.10: 75398758-75398758 on Build GRCh38
rs777654355 Chr.10: 75398742-75398742 on Build GRCh38
rs935161441 Chr.10: 75398712-75398712 on Build GRCh38
rs553682138 Chr.10: 75398743-75398743 on Build GRCh38
rs768792308 Chr.10: 75398782-75398782 on Build GRCh38
rs974754880 Chr.10: 75398722-75398722 on Build GRCh38

View Full Product Details