Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37885218-37885491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR87
Entrez Gene ID
83889
Gene Name
WD repeat domain 87
Gene Aliases
NYD-SP11
UniGene
Hs.664194
Gene Chromosome Location
Chr.19: 37884577-37906706 on Build GRCh38
Transcripts
NM_001291088.1 NM_031951.4 XM_011527360.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6439936 c.8202G>A p.W2734* Chr.19: 37885469-37885469 on Build GRCh38
7397722 c.8199C>A p.F2733L Chr.19: 37885472-37885472 on Build GRCh38
1734551 c.8276delA p.K2759fs*57 Chr.19: 37885395-37885395 on Build GRCh38
4510217 c.8422C>T p.Q2808* Chr.19: 37885249-37885249 on Build GRCh38
4510216 c.8305C>T p.Q2769* Chr.19: 37885249-37885249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752982361 Chr.19: 37885329-37885329 on Build GRCh38
rs927966832 Chr.19: 37885254-37885254 on Build GRCh38
rs755689018 Chr.19: 37885417-37885417 on Build GRCh38
rs943017090 Chr.19: 37885369-37885369 on Build GRCh38
rs982150812 Chr.19: 37885267-37885267 on Build GRCh38
rs761553527 Chr.19: 37885343-37885343 on Build GRCh38
rs1035008278 Chr.19: 37885268-37885268 on Build GRCh38
rs757306523 Chr.19: 37885387-37885387 on Build GRCh38
rs554726961 Chr.19: 37885289-37885289 on Build GRCh38
rs765235580 Chr.19: 37885320-37885320 on Build GRCh38
rs901498651 Chr.19: 37885426-37885426 on Build GRCh38
rs765398771 Chr.19: 37885269-37885269 on Build GRCh38
rs1049387071 Chr.19: 37885453-37885453 on Build GRCh38
rs745756486 Chr.19: 37885402-37885402 on Build GRCh38
rs117520417 Chr.19: 37885345-37885345 on Build GRCh38
rs946919831 Chr.19: 37885330-37885330 on Build GRCh38
rs544946317 Chr.19: 37885436-37885436 on Build GRCh38
rs760029944 Chr.19: 37885260-37885260 on Build GRCh38
rs959364670 Chr.19: 37885281-37885281 on Build GRCh38
rs866427542 Chr.19: 37885449-37885449 on Build GRCh38
rs200596982 Chr.19: 37885344-37885344 on Build GRCh38
rs73027454 Chr.19: 37885463-37885463 on Build GRCh38
rs993273559 Chr.19: 37885445-37885445 on Build GRCh38
rs577492453 Chr.19: 37885431-37885431 on Build GRCh38
rs185291608 Chr.19: 37885452-37885452 on Build GRCh38
rs766850781 Chr.19: 37885255-37885255 on Build GRCh38
rs374652986 Chr.19: 37885389-37885389 on Build GRCh38
rs367616075 Chr.19: 37885394-37885394 on Build GRCh38
rs754296189 Chr.19: 37885277-37885277 on Build GRCh38
rs763670486 Chr.19: 37885394-37885395 on Build GRCh38
rs991067154 Chr.19: 37885313-37885313 on Build GRCh38

View Full Product Details