Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32543975-32544226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOPORS
Entrez Gene ID
10210
Gene Name
TOP1 binding arginine/serine rich protein
Gene Aliases
LUN, P53BP3, RP31, TP53BPL
UniGene
Hs.589962
Gene Chromosome Location
Chr.9: 32540544-32552628 on Build GRCh38
Transcripts
NM_001195622.1 NM_005802.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1108199 c.365A>C p.K122T Chr.9: 32544160-32544160 on Build GRCh38
2772140 c.329T>C p.F110S Chr.9: 32544196-32544196 on Build GRCh38
6219445 c.542G>A p.R181H Chr.9: 32543983-32543983 on Build GRCh38
3906827 c.517A>G p.T173A Chr.9: 32544008-32544008 on Build GRCh38
1636154 c.442A>G p.I148V Chr.9: 32544083-32544083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776603944 Chr.9: 32544007-32544007 on Build GRCh38
rs763251504 Chr.9: 32544045-32544045 on Build GRCh38
rs143962770 Chr.9: 32543999-32543999 on Build GRCh38
rs757491435 Chr.9: 32544149-32544149 on Build GRCh38
rs777936936 Chr.9: 32544148-32544148 on Build GRCh38
rs755458435 Chr.9: 32544108-32544108 on Build GRCh38
rs767016930 Chr.9: 32544083-32544083 on Build GRCh38
rs769631149 Chr.9: 32544015-32544015 on Build GRCh38
rs771036401 Chr.9: 32544005-32544005 on Build GRCh38
rs745939345 Chr.9: 32544171-32544171 on Build GRCh38
rs758807913 Chr.9: 32544144-32544144 on Build GRCh38
rs200690745 Chr.9: 32544027-32544027 on Build GRCh38
rs778023328 Chr.9: 32543998-32543998 on Build GRCh38
rs779913865 Chr.9: 32544175-32544175 on Build GRCh38
rs200405067 Chr.9: 32544173-32544173 on Build GRCh38
rs753042372 Chr.9: 32544142-32544142 on Build GRCh38
rs17855104 Chr.9: 32544065-32544065 on Build GRCh38
rs765671154 Chr.9: 32544139-32544139 on Build GRCh38
rs749070735 Chr.9: 32544193-32544193 on Build GRCh38
rs763826828 Chr.9: 32544028-32544028 on Build GRCh38
rs749926750 Chr.9: 32544105-32544105 on Build GRCh38
rs375839984 Chr.9: 32544019-32544019 on Build GRCh38
rs879157854 Chr.9: 32544117-32544117 on Build GRCh38
rs147302494 Chr.9: 32544165-32544165 on Build GRCh38
rs761378644 Chr.9: 32544054-32544054 on Build GRCh38
rs745768297 Chr.9: 32544012-32544012 on Build GRCh38

View Full Product Details