Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17814504-17814757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SREBF1
Entrez Gene ID
6720
Gene Name
sterol regulatory element binding transcription factor 1
Gene Aliases
SREBP-1c, SREBP1, SREBP1a, bHLHd1
UniGene
Hs.592123
Gene Chromosome Location
Chr.17: 17811349-17837017 on Build GRCh38
Transcripts
NM_001005291.2 NM_001321096.2 NM_004176.4 XM_005256772.4 XM_017024970.1 XM_017024971.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5981158 c.2681C>T p.A894V Chr.17: 17814669-17814669 on Build GRCh38
3691396 c.2806C>T p.R936W Chr.17: 17814634-17814634 on Build GRCh38
3691397 c.2716C>T p.R906W Chr.17: 17814634-17814634 on Build GRCh38
5987225 c.2702T>A p.V901E Chr.17: 17814648-17814648 on Build GRCh38
4729969 c.2774A>G p.E925G Chr.17: 17814666-17814666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199944765 Chr.17: 17814581-17814581 on Build GRCh38
rs923223365 Chr.17: 17814715-17814715 on Build GRCh38
rs368912504 Chr.17: 17814642-17814642 on Build GRCh38
rs778157990 Chr.17: 17814580-17814580 on Build GRCh38
rs529460681 Chr.17: 17814687-17814687 on Build GRCh38
rs564110856 Chr.17: 17814571-17814571 on Build GRCh38
rs375637668 Chr.17: 17814585-17814585 on Build GRCh38
rs373272335 Chr.17: 17814738-17814738 on Build GRCh38
rs576886028 Chr.17: 17814669-17814669 on Build GRCh38
rs767791838 Chr.17: 17814576-17814576 on Build GRCh38
rs36027058 Chr.17: 17814637-17814638 on Build GRCh38
rs757078927 Chr.17: 17814688-17814688 on Build GRCh38
rs939900086 Chr.17: 17814641-17814641 on Build GRCh38
rs1035467089 Chr.17: 17814729-17814729 on Build GRCh38
rs952807362 Chr.17: 17814640-17814640 on Build GRCh38
rs1036260746 Chr.17: 17814536-17814536 on Build GRCh38
rs372600642 Chr.17: 17814539-17814539 on Build GRCh38
rs773687594 Chr.17: 17814664-17814664 on Build GRCh38
rs775182965 Chr.17: 17814654-17814654 on Build GRCh38
rs892700133 Chr.17: 17814573-17814573 on Build GRCh38
rs888869620 Chr.17: 17814562-17814562 on Build GRCh38
rs764217259 Chr.17: 17814684-17814684 on Build GRCh38
rs371883515 Chr.17: 17814604-17814604 on Build GRCh38
rs933401209 Chr.17: 17814616-17814616 on Build GRCh38
rs560879600 Chr.17: 17814674-17814674 on Build GRCh38
rs56088670 Chr.17: 17814677-17814677 on Build GRCh38
rs543156769 Chr.17: 17814698-17814698 on Build GRCh38
rs749283714 Chr.17: 17814633-17814633 on Build GRCh38
rs980798433 Chr.17: 17814605-17814605 on Build GRCh38
rs745682611 Chr.17: 17814653-17814653 on Build GRCh38
rs375856719 Chr.17: 17814635-17814635 on Build GRCh38
rs202180542 Chr.17: 17814737-17814737 on Build GRCh38
rs778925742 Chr.17: 17814718-17814718 on Build GRCh38
rs1000978803 Chr.17: 17814565-17814565 on Build GRCh38
rs921937534 Chr.17: 17814612-17814612 on Build GRCh38
rs746518916 Chr.17: 17814656-17814656 on Build GRCh38
rs914856170 Chr.17: 17814572-17814572 on Build GRCh38
rs760812071 Chr.17: 17814679-17814679 on Build GRCh38
rs909070855 Chr.17: 17814666-17814666 on Build GRCh38
rs768195955 Chr.17: 17814663-17814663 on Build GRCh38
rs1051897662 Chr.17: 17814629-17814629 on Build GRCh38
rs771472597 Chr.17: 17814671-17814671 on Build GRCh38

View Full Product Details