Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183666885-183667110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC11
Entrez Gene ID
60684
Gene Name
trafficking protein particle complex 11
Gene Aliases
C4orf41, FOIGR, GRY, LGMD2S
UniGene
Hs.443240
Gene Chromosome Location
Chr.4: 183659267-183713594 on Build GRCh38
Transcripts
NM_021942.5 NM_199053.2 XM_017008537.1 XM_017008538.1 XM_017008539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6469438 c.392G>A p.R131K Chr.4: 183667077-183667077 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs926392732 Chr.4: 183667059-183667059 on Build GRCh38
rs774078744 Chr.4: 183667084-183667084 on Build GRCh38
rs946388636 Chr.4: 183666996-183666996 on Build GRCh38
rs566450197 Chr.4: 183667081-183667081 on Build GRCh38
rs749273905 Chr.4: 183667047-183667047 on Build GRCh38
rs897945331 Chr.4: 183667043-183667043 on Build GRCh38
rs4282227 Chr.4: 183666968-183666968 on Build GRCh38
rs555311904 Chr.4: 183666944-183666944 on Build GRCh38
rs765501476 Chr.4: 183667046-183667046 on Build GRCh38
rs78001580 Chr.4: 183666995-183666995 on Build GRCh38
rs937974071 Chr.4: 183666929-183666929 on Build GRCh38
rs535093370 Chr.4: 183666919-183666919 on Build GRCh38
rs780488025 Chr.4: 183667062-183667062 on Build GRCh38
rs534647765 Chr.4: 183666927-183666927 on Build GRCh38
rs758790469 Chr.4: 183667053-183667053 on Build GRCh38
rs369613803 Chr.4: 183667069-183667069 on Build GRCh38
rs757576636 Chr.4: 183667036-183667036 on Build GRCh38
rs577784957 Chr.4: 183666993-183666993 on Build GRCh38
rs1045155771 Chr.4: 183667010-183667010 on Build GRCh38
rs750824619 Chr.4: 183667052-183667052 on Build GRCh38

View Full Product Details