Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10260285-10260540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TATDN2
Entrez Gene ID
9797
Gene Name
TatD DNase domain containing 2
Gene Aliases
-
UniGene
Hs.475401
Gene Chromosome Location
Chr.3: 10248493-10281222 on Build GRCh38
Transcripts
NM_014760.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5021366 c.650A>G p.H217R Chr.3: 10260372-10260372 on Build GRCh38
5904776 c.766G>A p.V256I Chr.3: 10260488-10260488 on Build GRCh38
4583754 c.640G>A p.A214T Chr.3: 10260362-10260362 on Build GRCh38
2916090 c.594G>A p.S198S Chr.3: 10260316-10260316 on Build GRCh38
2916092 c.616G>A p.A206T Chr.3: 10260338-10260338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750296145 Chr.3: 10260396-10260396 on Build GRCh38
rs371526622 Chr.3: 10260322-10260322 on Build GRCh38
rs763845193 Chr.3: 10260380-10260380 on Build GRCh38
rs2241313 Chr.3: 10260361-10260361 on Build GRCh38
rs781503431 Chr.3: 10260494-10260499 on Build GRCh38
rs781382002 Chr.3: 10260436-10260436 on Build GRCh38
rs753432686 Chr.3: 10260323-10260323 on Build GRCh38
rs748472416 Chr.3: 10260439-10260439 on Build GRCh38
rs559746296 Chr.3: 10260387-10260387 on Build GRCh38
rs1029735291 Chr.3: 10260491-10260491 on Build GRCh38
rs761566077 Chr.3: 10260494-10260494 on Build GRCh38
rs748227821 Chr.3: 10260362-10260362 on Build GRCh38
rs749968523 Chr.3: 10260505-10260505 on Build GRCh38
rs374555335 Chr.3: 10260326-10260326 on Build GRCh38
rs370249364 Chr.3: 10260410-10260410 on Build GRCh38
rs768051337 Chr.3: 10260483-10260483 on Build GRCh38
rs367785698 Chr.3: 10260369-10260369 on Build GRCh38
rs779967912 Chr.3: 10260500-10260500 on Build GRCh38
rs758122936 Chr.3: 10260338-10260338 on Build GRCh38
rs765691630 Chr.3: 10260305-10260305 on Build GRCh38
rs141238234 Chr.3: 10260476-10260476 on Build GRCh38
rs199748216 Chr.3: 10260351-10260351 on Build GRCh38
rs557554443 Chr.3: 10260337-10260337 on Build GRCh38
rs773394766 Chr.3: 10260461-10260461 on Build GRCh38
rs755881321 Chr.3: 10260320-10260320 on Build GRCh38
rs758405905 Chr.3: 10260397-10260397 on Build GRCh38
rs954210878 Chr.3: 10260493-10260493 on Build GRCh38
rs376953395 Chr.3: 10260310-10260310 on Build GRCh38
rs2241314 Chr.3: 10260372-10260372 on Build GRCh38
rs559267711 Chr.3: 10260478-10260478 on Build GRCh38
rs394558 Chr.3: 10260488-10260488 on Build GRCh38
rs749710932 Chr.3: 10260474-10260474 on Build GRCh38
rs769976539 Chr.3: 10260368-10260368 on Build GRCh38
rs779771233 Chr.3: 10260400-10260400 on Build GRCh38
rs760366340 Chr.3: 10260373-10260373 on Build GRCh38
rs755123581 Chr.3: 10260422-10260422 on Build GRCh38
rs376639930 Chr.3: 10260316-10260316 on Build GRCh38
rs747052958 Chr.3: 10260386-10260403 on Build GRCh38
rs751295525 Chr.3: 10260513-10260513 on Build GRCh38
rs148251801 Chr.3: 10260475-10260475 on Build GRCh38
rs766306350 Chr.3: 10260507-10260507 on Build GRCh38
rs746835629 Chr.3: 10260418-10260418 on Build GRCh38
rs868059814 Chr.3: 10260321-10260321 on Build GRCh38
rs779863358 Chr.3: 10260347-10260347 on Build GRCh38
rs529268928 Chr.3: 10260371-10260371 on Build GRCh38
rs747207064 Chr.3: 10260348-10260348 on Build GRCh38
rs778728614 Chr.3: 10260331-10260331 on Build GRCh38
rs1004752505 Chr.3: 10260466-10260466 on Build GRCh38
rs141030272 Chr.3: 10260447-10260447 on Build GRCh38
rs762755485 Chr.3: 10260506-10260506 on Build GRCh38
rs750744085 Chr.3: 10260306-10260306 on Build GRCh38
rs147562335 Chr.3: 10260392-10260392 on Build GRCh38
rs755304679 Chr.3: 10260404-10260424 on Build GRCh38
rs949807237 Chr.3: 10260304-10260304 on Build GRCh38
rs374060667 Chr.3: 10260308-10260308 on Build GRCh38
rs764868389 Chr.3: 10260393-10260393 on Build GRCh38

View Full Product Details