Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36626946-36627209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF382
Entrez Gene ID
84911
Gene Name
zinc finger protein 382
Gene Aliases
KS1
UniGene
Hs.631591
Gene Chromosome Location
Chr.19: 36605305-36628597 on Build GRCh38
Transcripts
NM_001256838.1 NM_032825.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1233962 c.1129G>A p.E377K Chr.19: 36627026-36627026 on Build GRCh38
1750843 c.1105A>T p.R369* Chr.19: 36627002-36627002 on Build GRCh38
5024530 c.1179C>A p.Y393* Chr.19: 36627079-36627079 on Build GRCh38
6829724 c.1139A>G p.Y380C Chr.19: 36627036-36627036 on Build GRCh38
126381 c.1286C>A p.T429N Chr.19: 36627183-36627183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955316468 Chr.19: 36627106-36627106 on Build GRCh38
rs1005527700 Chr.19: 36627124-36627124 on Build GRCh38
rs975309679 Chr.19: 36627054-36627054 on Build GRCh38
rs776479508 Chr.19: 36627048-36627048 on Build GRCh38
rs765710784 Chr.19: 36627166-36627166 on Build GRCh38
rs568283174 Chr.19: 36627123-36627123 on Build GRCh38
rs769162959 Chr.19: 36627137-36627137 on Build GRCh38
rs772376239 Chr.19: 36627163-36627163 on Build GRCh38
rs537671705 Chr.19: 36627000-36627000 on Build GRCh38
rs749359129 Chr.19: 36627005-36627005 on Build GRCh38
rs148103215 Chr.19: 36627022-36627022 on Build GRCh38
rs746247001 Chr.19: 36626995-36626995 on Build GRCh38
rs773090722 Chr.19: 36627024-36627024 on Build GRCh38
rs757614823 Chr.19: 36626982-36626982 on Build GRCh38
rs201684196 Chr.19: 36627079-36627079 on Build GRCh38
rs779075034 Chr.19: 36627145-36627145 on Build GRCh38
rs781469678 Chr.19: 36626989-36626989 on Build GRCh38
rs4439880 Chr.19: 36627096-36627096 on Build GRCh38
rs780590866 Chr.19: 36627118-36627118 on Build GRCh38
rs750747548 Chr.19: 36627099-36627099 on Build GRCh38
rs766307873 Chr.19: 36627045-36627045 on Build GRCh38
rs894031247 Chr.19: 36627033-36627033 on Build GRCh38
rs906739724 Chr.19: 36627084-36627084 on Build GRCh38
rs1032659383 Chr.19: 36626999-36626999 on Build GRCh38
rs1045241896 Chr.19: 36627073-36627073 on Build GRCh38
rs757988766 Chr.19: 36627094-36627094 on Build GRCh38
rs748257565 Chr.19: 36627155-36627155 on Build GRCh38
rs752404502 Chr.19: 36626981-36626981 on Build GRCh38
rs1025700489 Chr.19: 36627097-36627097 on Build GRCh38
rs200498882 Chr.19: 36627034-36627034 on Build GRCh38
rs201197350 Chr.19: 36627082-36627082 on Build GRCh38
rs185047483 Chr.19: 36626992-36626992 on Build GRCh38
rs577820635 Chr.19: 36627059-36627059 on Build GRCh38
rs1003622333 Chr.19: 36627087-36627087 on Build GRCh38
rs768671768 Chr.19: 36627019-36627019 on Build GRCh38
rs867969154 Chr.19: 36627057-36627057 on Build GRCh38
rs756566547 Chr.19: 36626998-36626998 on Build GRCh38
rs935644765 Chr.19: 36627014-36627014 on Build GRCh38
rs760133178 Chr.19: 36627117-36627117 on Build GRCh38
rs553797186 Chr.19: 36627127-36627127 on Build GRCh38

View Full Product Details