Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90258542-90258793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIB1
Entrez Gene ID
10519
Gene Name
calcium and integrin binding 1
Gene Aliases
CIB, CIBP, KIP1, PRKDCIP, SIP2-28
UniGene
Hs.715556
Gene Chromosome Location
Chr.15: 90230245-90265759 on Build GRCh38
Transcripts
NR_102428.1 NR_104604.1

Gene Symbol
TTLL13P
Entrez Gene ID
440307
Gene Name
tubulin tyrosine ligase like 13, pseudogene
Gene Aliases
TTLL13
UniGene
Hs.632164
Gene Chromosome Location
Chr.15: 90249532-90259088 on Build GRCh38
Transcripts
NR_102428.1 NR_104604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4829008 c.1199C>G p.S400* Chr.15: 90258774-90258774 on Build GRCh38
1678681 c.1213G>T p.E405* Chr.15: 90258788-90258788 on Build GRCh38
1678682 c.1213G>T p.E405* Chr.15: 90258788-90258788 on Build GRCh38
4829007 c.1199C>G p.S400* Chr.15: 90258774-90258774 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs141146113 Chr.15: 90258632-90258632 on Build GRCh38
rs574235900 Chr.15: 90258738-90258738 on Build GRCh38
rs753796405 Chr.15: 90258736-90258736 on Build GRCh38
rs566840658 Chr.15: 90258688-90258688 on Build GRCh38
rs781411688 Chr.15: 90258758-90258758 on Build GRCh38
rs143952686 Chr.15: 90258768-90258768 on Build GRCh38
rs765629027 Chr.15: 90258719-90258719 on Build GRCh38
rs752129471 Chr.15: 90258621-90258621 on Build GRCh38
rs569738033 Chr.15: 90258664-90258664 on Build GRCh38
rs965581942 Chr.15: 90258587-90258587 on Build GRCh38
rs146894802 Chr.15: 90258676-90258676 on Build GRCh38
rs551293216 Chr.15: 90258661-90258661 on Build GRCh38
rs371779864 Chr.15: 90258726-90258726 on Build GRCh38
rs754928759 Chr.15: 90258659-90258659 on Build GRCh38
rs888813438 Chr.15: 90258720-90258720 on Build GRCh38
rs769018866 Chr.15: 90258711-90258711 on Build GRCh38
rs778872355 Chr.15: 90258766-90258766 on Build GRCh38
rs757479674 Chr.15: 90258754-90258754 on Build GRCh38
rs544425042 Chr.15: 90258759-90258759 on Build GRCh38
rs369297616 Chr.15: 90258705-90258705 on Build GRCh38
rs754938455 Chr.15: 90258765-90258765 on Build GRCh38
rs375075471 Chr.15: 90258769-90258769 on Build GRCh38
rs975599690 Chr.15: 90258631-90258631 on Build GRCh38
rs751738285 Chr.15: 90258753-90258753 on Build GRCh38
rs770420760 Chr.15: 90258692-90258692 on Build GRCh38
rs763326678 Chr.15: 90258733-90258733 on Build GRCh38
rs369004061 Chr.15: 90258643-90258643 on Build GRCh38
rs760014002 Chr.15: 90258718-90258718 on Build GRCh38

View Full Product Details