Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 28682116-28682351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAMBI
Entrez Gene ID
25805
Gene Name
BMP and activin membrane bound inhibitor
Gene Aliases
NMA
UniGene
Hs.533336
Gene Chromosome Location
Chr.10: 28677495-28682939 on Build GRCh38
Transcripts
NM_012342.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6065901 c.514G>T p.A172S Chr.10: 28682132-28682132 on Build GRCh38
3741631 c.728T>A p.L243H Chr.10: 28682346-28682346 on Build GRCh38
7113453 c.655G>A p.V219I Chr.10: 28682273-28682273 on Build GRCh38
3437106 c.646A>C p.M216L Chr.10: 28682264-28682264 on Build GRCh38
342753 c.615G>T p.K205N Chr.10: 28682233-28682233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771656990 Chr.10: 28682193-28682196 on Build GRCh38
rs975499185 Chr.10: 28682190-28682190 on Build GRCh38
rs764485833 Chr.10: 28682286-28682286 on Build GRCh38
rs573231648 Chr.10: 28682271-28682271 on Build GRCh38
rs759754106 Chr.10: 28682194-28682194 on Build GRCh38
rs201458451 Chr.10: 28682178-28682178 on Build GRCh38
rs767932179 Chr.10: 28682195-28682195 on Build GRCh38
rs146850430 Chr.10: 28682256-28682256 on Build GRCh38
rs539221766 Chr.10: 28682220-28682220 on Build GRCh38
rs766000717 Chr.10: 28682303-28682303 on Build GRCh38
rs200922139 Chr.10: 28682215-28682215 on Build GRCh38
rs765591573 Chr.10: 28682285-28682285 on Build GRCh38
rs540765877 Chr.10: 28682302-28682302 on Build GRCh38
rs989917730 Chr.10: 28682263-28682263 on Build GRCh38
rs752372665 Chr.10: 28682323-28682323 on Build GRCh38
rs72809669 Chr.10: 28682240-28682240 on Build GRCh38
rs778966773 Chr.10: 28682156-28682157 on Build GRCh38
rs575182425 Chr.10: 28682196-28682196 on Build GRCh38
rs763221755 Chr.10: 28682316-28682317 on Build GRCh38
rs965367198 Chr.10: 28682318-28682318 on Build GRCh38
rs998923608 Chr.10: 28682277-28682277 on Build GRCh38
rs949321109 Chr.10: 28682223-28682223 on Build GRCh38
rs748105995 Chr.10: 28682176-28682176 on Build GRCh38
rs377447657 Chr.10: 28682309-28682309 on Build GRCh38
rs749221213 Chr.10: 28682261-28682261 on Build GRCh38
rs747909338 Chr.10: 28682238-28682238 on Build GRCh38
rs146161229 Chr.10: 28682172-28682172 on Build GRCh38
rs754616555 Chr.10: 28682237-28682237 on Build GRCh38
rs781462756 Chr.10: 28682301-28682301 on Build GRCh38
rs1012119670 Chr.10: 28682290-28682290 on Build GRCh38
rs373511407 Chr.10: 28682207-28682207 on Build GRCh38
rs556798784 Chr.10: 28682179-28682179 on Build GRCh38
rs754706436 Chr.10: 28682314-28682314 on Build GRCh38
rs762890643 Chr.10: 28682182-28682182 on Build GRCh38
rs568094718 Chr.10: 28682169-28682169 on Build GRCh38
rs369743386 Chr.10: 28682148-28682148 on Build GRCh38
rs762204835 Chr.10: 28682275-28682275 on Build GRCh38
rs776403208 Chr.10: 28682228-28682228 on Build GRCh38
rs771023057 Chr.10: 28682187-28682187 on Build GRCh38
rs924524101 Chr.10: 28682235-28682235 on Build GRCh38
rs772877740 Chr.10: 28682198-28682203 on Build GRCh38
rs746602206 Chr.10: 28682200-28682201 on Build GRCh38
rs983183120 Chr.10: 28682145-28682145 on Build GRCh38
rs1046146756 Chr.10: 28682327-28682327 on Build GRCh38
rs140192557 Chr.10: 28682233-28682233 on Build GRCh38
rs758102766 Chr.10: 28682141-28682141 on Build GRCh38
rs779610447 Chr.10: 28682232-28682232 on Build GRCh38
rs373310257 Chr.10: 28682272-28682272 on Build GRCh38
rs746628144 Chr.10: 28682166-28682166 on Build GRCh38
rs546258370 Chr.10: 28682151-28682151 on Build GRCh38
rs774598560 Chr.10: 28682191-28682191 on Build GRCh38
rs748148816 Chr.10: 28682176-28682177 on Build GRCh38
rs561923649 Chr.10: 28682320-28682320 on Build GRCh38
rs369100188 Chr.10: 28682264-28682264 on Build GRCh38
rs148416941 Chr.10: 28682292-28682292 on Build GRCh38
rs377723568 Chr.10: 28682284-28682284 on Build GRCh38
rs751315477 Chr.10: 28682236-28682236 on Build GRCh38
rs1029297308 Chr.10: 28682205-28682205 on Build GRCh38
rs770499180 Chr.10: 28682205-28682213 on Build GRCh38

View Full Product Details