Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30335616-30335849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM39
Entrez Gene ID
56658
Gene Name
tripartite motif containing 39
Gene Aliases
RNF23, TFP, TRIM39B
UniGene
Hs.413493
Gene Chromosome Location
Chr.6: 30326844-30343729 on Build GRCh38
Transcripts
NM_021253.3 NM_172016.2 NM_001199119.1

Gene Symbol
TRIM39-RPP21
Entrez Gene ID
202658
Gene Name
TRIM39-RPP21 readthrough
Gene Aliases
TRIM39R
UniGene
Hs.413493
Gene Chromosome Location
Chr.6: 30329311-30346858 on Build GRCh38
Transcripts
NM_021253.3 NM_172016.2 NM_001199119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3251262 c.566G>A p.R189H Chr.6: 30335761-30335761 on Build GRCh38
4160553 c.565C>A p.R189S Chr.6: 30335760-30335760 on Build GRCh38
4856891 c.616G>A p.E206K Chr.6: 30335811-30335811 on Build GRCh38
3394046 c.640C>T p.R214* Chr.6: 30335835-30335835 on Build GRCh38
3394048 c.640C>T p.R214* Chr.6: 30335835-30335835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752461560 Chr.6: 30335813-30335813 on Build GRCh38
rs973243732 Chr.6: 30335723-30335723 on Build GRCh38
rs551694859 Chr.6: 30335752-30335752 on Build GRCh38
rs867626538 Chr.6: 30335781-30335781 on Build GRCh38
rs965283260 Chr.6: 30335677-30335677 on Build GRCh38
rs776895166 Chr.6: 30335734-30335734 on Build GRCh38
rs766537366 Chr.6: 30335749-30335749 on Build GRCh38
rs772415111 Chr.6: 30335703-30335703 on Build GRCh38
rs1015008870 Chr.6: 30335680-30335680 on Build GRCh38
rs1017770623 Chr.6: 30335655-30335655 on Build GRCh38
rs369381009 Chr.6: 30335700-30335700 on Build GRCh38
rs764875170 Chr.6: 30335803-30335803 on Build GRCh38
rs760707686 Chr.6: 30335790-30335790 on Build GRCh38
rs747609910 Chr.6: 30335728-30335728 on Build GRCh38
rs773973854 Chr.6: 30335824-30335824 on Build GRCh38
rs754429118 Chr.6: 30335802-30335802 on Build GRCh38
rs753367826 Chr.6: 30335797-30335797 on Build GRCh38
rs775785915 Chr.6: 30335760-30335760 on Build GRCh38
rs937864936 Chr.6: 30335816-30335816 on Build GRCh38
rs759652467 Chr.6: 30335740-30335740 on Build GRCh38
rs763079283 Chr.6: 30335761-30335761 on Build GRCh38
rs186760270 Chr.6: 30335731-30335731 on Build GRCh38
rs145694007 Chr.6: 30335776-30335776 on Build GRCh38
rs566267140 Chr.6: 30335718-30335718 on Build GRCh38
rs767399572 Chr.6: 30335764-30335764 on Build GRCh38
rs537592730 Chr.6: 30335795-30335795 on Build GRCh38
rs917248937 Chr.6: 30335765-30335765 on Build GRCh38

View Full Product Details