Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47846179-47846436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF81
Entrez Gene ID
347344
Gene Name
zinc finger protein 81
Gene Aliases
HFZ20, MRX45, dJ54B20.6
UniGene
Hs.114246
Gene Chromosome Location
Chr.X: 47836864-47925626 on Build GRCh38
Transcripts
NM_007137.3 XM_005272600.3 XM_011543899.2 XM_011543900.2 XM_017029486.1 XM_017029487.1

SNP Detail

SNP ID Chromosome Location
rs145478020 Chr.X: 47846285-47846285 on Build GRCh38
rs782490338 Chr.X: 47846252-47846252 on Build GRCh38
rs888393643 Chr.X: 47846280-47846280 on Build GRCh38
rs782275384 Chr.X: 47846213-47846213 on Build GRCh38
rs781959530 Chr.X: 47846301-47846301 on Build GRCh38
rs183846665 Chr.X: 47846275-47846275 on Build GRCh38
rs374414966 Chr.X: 47846261-47846261 on Build GRCh38
rs376023206 Chr.X: 47846279-47846279 on Build GRCh38
rs980705670 Chr.X: 47846402-47846402 on Build GRCh38
rs782804886 Chr.X: 47846352-47846352 on Build GRCh38
rs782314134 Chr.X: 47846318-47846318 on Build GRCh38
rs375317615 Chr.X: 47846335-47846335 on Build GRCh38
rs961035108 Chr.X: 47846317-47846317 on Build GRCh38
rs782316402 Chr.X: 47846267-47846267 on Build GRCh38
rs782229859 Chr.X: 47846383-47846383 on Build GRCh38
rs781857518 Chr.X: 47846236-47846236 on Build GRCh38
rs1013929536 Chr.X: 47846331-47846331 on Build GRCh38
rs997105196 Chr.X: 47846268-47846268 on Build GRCh38
rs371476881 Chr.X: 47846200-47846200 on Build GRCh38
rs782077493 Chr.X: 47846315-47846315 on Build GRCh38
rs782748474 Chr.X: 47846369-47846369 on Build GRCh38
rs782653647 Chr.X: 47846217-47846217 on Build GRCh38
rs782160408 Chr.X: 47846341-47846341 on Build GRCh38
rs782120231 Chr.X: 47846364-47846364 on Build GRCh38
rs782609524 Chr.X: 47846258-47846258 on Build GRCh38
rs782001834 Chr.X: 47846363-47846363 on Build GRCh38
rs782369848 Chr.X: 47846296-47846296 on Build GRCh38
rs1015172921 Chr.X: 47846286-47846286 on Build GRCh38
rs782473354 Chr.X: 47846236-47846236 on Build GRCh38

View Full Product Details