Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 68876903-68877158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN1
Entrez Gene ID
87
Gene Name
actinin alpha 1
Gene Aliases
BDPLT15
UniGene
Hs.235750
Gene Chromosome Location
Chr.14: 68874123-68979366 on Build GRCh38
Transcripts
NM_001102.3 NM_001130004.1 NM_001130005.1 XM_011537265.2 XM_011537266.2 XM_011537267.2 XM_011537268.2 XM_017021720.1 XM_017021721.1 XM_017021722.1 XM_017021723.1 XM_017021725.1 XM_017021726.1 XM_017021727.1 XM_017021728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4481530 c.2518C>T p.R840C Chr.14: 68877150-68877150 on Build GRCh38
4872085 c.2580G>A p.G860G Chr.14: 68877088-68877088 on Build GRCh38
4341108 c.2529C>T p.A843A Chr.14: 68877139-68877139 on Build GRCh38
5571913 c.2487C>A p.V829V Chr.14: 68877115-68877115 on Build GRCh38
5571912 c.2553C>A p.V851V Chr.14: 68877115-68877115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775542710 Chr.14: 68877096-68877096 on Build GRCh38
rs200585276 Chr.14: 68877035-68877035 on Build GRCh38
rs745643096 Chr.14: 68877083-68877083 on Build GRCh38
rs1031556481 Chr.14: 68877001-68877001 on Build GRCh38
rs540315666 Chr.14: 68876942-68876942 on Build GRCh38
rs1041029000 Chr.14: 68877066-68877066 on Build GRCh38
rs989647975 Chr.14: 68877015-68877015 on Build GRCh38
rs768821359 Chr.14: 68877138-68877138 on Build GRCh38
rs1018791199 Chr.14: 68876948-68876948 on Build GRCh38
rs774599678 Chr.14: 68877139-68877139 on Build GRCh38
rs75219984 Chr.14: 68876987-68876987 on Build GRCh38
rs966246414 Chr.14: 68876956-68876956 on Build GRCh38
rs754400788 Chr.14: 68877042-68877042 on Build GRCh38
rs757882380 Chr.14: 68877052-68877052 on Build GRCh38
rs201687580 Chr.14: 68877091-68877091 on Build GRCh38
rs200162044 Chr.14: 68877063-68877063 on Build GRCh38
rs1803030 Chr.14: 68877126-68877126 on Build GRCh38
rs201843819 Chr.14: 68877101-68877101 on Build GRCh38
rs957274308 Chr.14: 68877009-68877009 on Build GRCh38
rs915551718 Chr.14: 68877050-68877050 on Build GRCh38
rs777295117 Chr.14: 68877055-68877055 on Build GRCh38
rs564964904 Chr.14: 68876968-68876968 on Build GRCh38
rs763019862 Chr.14: 68877109-68877109 on Build GRCh38
rs780971198 Chr.14: 68877079-68877079 on Build GRCh38

View Full Product Details