Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6640304-6640553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACRBP
Entrez Gene ID
84519
Gene Name
acrosin binding protein
Gene Aliases
CT23, OY-TES-1, SP32
UniGene
Hs.123239
Gene Chromosome Location
Chr.12: 6638076-6647414 on Build GRCh38
Transcripts
NM_032489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6199860 c.1166C>G p.A389G Chr.12: 6640434-6640434 on Build GRCh38
4044369 c.1209C>A p.P403P Chr.12: 6640391-6640391 on Build GRCh38
2098762 c.1179G>A p.R393R Chr.12: 6640421-6640421 on Build GRCh38
4044370 c.1189G>A p.D397N Chr.12: 6640411-6640411 on Build GRCh38
6234110 c.1170C>A p.S390R Chr.12: 6640430-6640430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767089367 Chr.12: 6640450-6640450 on Build GRCh38
rs866151180 Chr.12: 6640492-6640492 on Build GRCh38
rs903170718 Chr.12: 6640379-6640379 on Build GRCh38
rs758221974 Chr.12: 6640524-6640524 on Build GRCh38
rs771778088 Chr.12: 6640323-6640323 on Build GRCh38
rs763161133 Chr.12: 6640344-6640344 on Build GRCh38
rs915008302 Chr.12: 6640338-6640338 on Build GRCh38
rs200586817 Chr.12: 6640425-6640425 on Build GRCh38
rs573615966 Chr.12: 6640494-6640495 on Build GRCh38
rs150031699 Chr.12: 6640411-6640411 on Build GRCh38
rs761778569 Chr.12: 6640504-6640504 on Build GRCh38
rs764217842 Chr.12: 6640501-6640501 on Build GRCh38
rs965532966 Chr.12: 6640391-6640391 on Build GRCh38
rs774640239 Chr.12: 6640349-6640349 on Build GRCh38
rs778084989 Chr.12: 6640438-6640438 on Build GRCh38
rs1057234968 Chr.12: 6640531-6640531 on Build GRCh38
rs376661923 Chr.12: 6640423-6640423 on Build GRCh38
rs750168129 Chr.12: 6640522-6640522 on Build GRCh38
rs947283301 Chr.12: 6640350-6640350 on Build GRCh38
rs768217426 Chr.12: 6640427-6640427 on Build GRCh38
rs149157588 Chr.12: 6640482-6640482 on Build GRCh38
rs372213716 Chr.12: 6640514-6640514 on Build GRCh38
rs371593728 Chr.12: 6640332-6640332 on Build GRCh38
rs201606879 Chr.12: 6640331-6640331 on Build GRCh38
rs145271222 Chr.12: 6640412-6640412 on Build GRCh38
rs750146719 Chr.12: 6640481-6640481 on Build GRCh38
rs199911393 Chr.12: 6640502-6640502 on Build GRCh38
rs771412740 Chr.12: 6640460-6640460 on Build GRCh38
rs778764004 Chr.12: 6640401-6640401 on Build GRCh38
rs779617398 Chr.12: 6640527-6640527 on Build GRCh38
rs760775318 Chr.12: 6640500-6640500 on Build GRCh38
rs369836113 Chr.12: 6640480-6640480 on Build GRCh38
rs368787320 Chr.12: 6640387-6640387 on Build GRCh38
rs757195399 Chr.12: 6640398-6640398 on Build GRCh38
rs953143489 Chr.12: 6640451-6640451 on Build GRCh38
rs577478690 Chr.12: 6640351-6640351 on Build GRCh38
rs766780265 Chr.12: 6640345-6640345 on Build GRCh38
rs749793254 Chr.12: 6640440-6640440 on Build GRCh38
rs1030521206 Chr.12: 6640415-6640415 on Build GRCh38
rs752805695 Chr.12: 6640352-6640352 on Build GRCh38
rs763868441 Chr.12: 6640372-6640372 on Build GRCh38
rs774750000 Chr.12: 6640464-6640464 on Build GRCh38
rs760865391 Chr.12: 6640354-6640354 on Build GRCh38
rs373672314 Chr.12: 6640376-6640376 on Build GRCh38
rs779701829 Chr.12: 6640422-6640422 on Build GRCh38
rs922988124 Chr.12: 6640330-6640330 on Build GRCh38

View Full Product Details