Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153775145-153775418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC27A3
Entrez Gene ID
11000
Gene Name
solute carrier family 27 member 3
Gene Aliases
ACSVL3, FATP3, VLCS-3
UniGene
Hs.438723
Gene Chromosome Location
Chr.1: 153774989-153780157 on Build GRCh38
Transcripts
NM_001317929.1 NM_024330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2208001 c.28C>T p.P10S Chr.1: 153775384-153775384 on Build GRCh38
6058058 c.16C>A p.R6S Chr.1: 153775372-153775372 on Build GRCh38
5477626 c.41A>T p.K14M Chr.1: 153775397-153775397 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756228690 Chr.1: 153775322-153775322 on Build GRCh38
rs776070776 Chr.1: 153775294-153775294 on Build GRCh38
rs185057219 Chr.1: 153775182-153775182 on Build GRCh38
rs539067087 Chr.1: 153775178-153775178 on Build GRCh38
rs573127426 Chr.1: 153775228-153775228 on Build GRCh38
rs779292108 Chr.1: 153775346-153775346 on Build GRCh38
rs759199864 Chr.1: 153775300-153775301 on Build GRCh38
rs1016626670 Chr.1: 153775248-153775248 on Build GRCh38
rs1016158589 Chr.1: 153775344-153775344 on Build GRCh38
rs966793831 Chr.1: 153775298-153775298 on Build GRCh38
rs542735734 Chr.1: 153775214-153775214 on Build GRCh38
rs377764357 Chr.1: 153775381-153775381 on Build GRCh38
rs770734530 Chr.1: 153775212-153775212 on Build GRCh38
rs774812411 Chr.1: 153775362-153775362 on Build GRCh38
rs549131824 Chr.1: 153775365-153775365 on Build GRCh38
rs780903625 Chr.1: 153775337-153775337 on Build GRCh38
rs778518509 Chr.1: 153775213-153775213 on Build GRCh38
rs768136591 Chr.1: 153775293-153775293 on Build GRCh38
rs745589728 Chr.1: 153775271-153775271 on Build GRCh38
rs930474655 Chr.1: 153775237-153775237 on Build GRCh38
rs771587570 Chr.1: 153775357-153775357 on Build GRCh38
rs755738692 Chr.1: 153775270-153775270 on Build GRCh38
rs767879824 Chr.1: 153775319-153775319 on Build GRCh38
rs777094679 Chr.1: 153775227-153775227 on Build GRCh38
rs900131274 Chr.1: 153775396-153775396 on Build GRCh38
rs751624762 Chr.1: 153775180-153775180 on Build GRCh38
rs112438704 Chr.1: 153775218-153775218 on Build GRCh38
rs367588342 Chr.1: 153775325-153775325 on Build GRCh38
rs779877530 Chr.1: 153775329-153775329 on Build GRCh38
rs1028583217 Chr.1: 153775281-153775281 on Build GRCh38
rs531156965 Chr.1: 153775341-153775341 on Build GRCh38
rs767393433 Chr.1: 153775340-153775341 on Build GRCh38
rs761418504 Chr.1: 153775296-153775296 on Build GRCh38
rs751808699 Chr.1: 153775312-153775312 on Build GRCh38
rs530584891 Chr.1: 153775351-153775351 on Build GRCh38
rs766335084 Chr.1: 153775385-153775385 on Build GRCh38
rs758162485 Chr.1: 153775327-153775327 on Build GRCh38
rs766888135 Chr.1: 153775309-153775309 on Build GRCh38
rs771915237 Chr.1: 153775274-153775274 on Build GRCh38
rs746829483 Chr.1: 153775334-153775334 on Build GRCh38
rs575288664 Chr.1: 153775198-153775198 on Build GRCh38
rs540566011 Chr.1: 153775263-153775263 on Build GRCh38
rs963294134 Chr.1: 153775364-153775364 on Build GRCh38
rs937469156 Chr.1: 153775297-153775297 on Build GRCh38
rs770135207 Chr.1: 153775299-153775299 on Build GRCh38
rs369246509 Chr.1: 153775262-153775262 on Build GRCh38
rs77171568 Chr.1: 153775326-153775326 on Build GRCh38
rs778911407 Chr.1: 153775265-153775265 on Build GRCh38
rs752650677 Chr.1: 153775321-153775321 on Build GRCh38
rs754585303 Chr.1: 153775336-153775336 on Build GRCh38
rs532703647 Chr.1: 153775276-153775276 on Build GRCh38
rs894899211 Chr.1: 153775199-153775200 on Build GRCh38
rs763381904 Chr.1: 153775260-153775260 on Build GRCh38
rs746708907 Chr.1: 153775290-153775290 on Build GRCh38
rs749277694 Chr.1: 153775211-153775211 on Build GRCh38
rs150756179 Chr.1: 153775371-153775371 on Build GRCh38
rs565610766 Chr.1: 153775264-153775264 on Build GRCh38
rs900181370 Chr.1: 153775267-153775267 on Build GRCh38
rs963179258 Chr.1: 153775262-153775262 on Build GRCh38
rs750715768 Chr.1: 153775252-153775252 on Build GRCh38
rs944445475 Chr.1: 153775377-153775377 on Build GRCh38
rs930592262 Chr.1: 153775333-153775333 on Build GRCh38
rs767787274 Chr.1: 153775368-153775368 on Build GRCh38
rs760167280 Chr.1: 153775226-153775226 on Build GRCh38
rs145256922 Chr.1: 153775380-153775380 on Build GRCh38
rs763221872 Chr.1: 153775304-153775304 on Build GRCh38
rs775836265 Chr.1: 153775369-153775369 on Build GRCh38
rs751289599 Chr.1: 153775387-153775387 on Build GRCh38
rs754783148 Chr.1: 153775393-153775393 on Build GRCh38
rs374424428 Chr.1: 153775353-153775353 on Build GRCh38
rs551251871 Chr.1: 153775302-153775302 on Build GRCh38

View Full Product Details