Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119154683-119154948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG4
Entrez Gene ID
64137
Gene Name
ATP binding cassette subfamily G member 4
Gene Aliases
WHITE2
UniGene
Hs.126378
Gene Chromosome Location
Chr.11: 119149040-119162666 on Build GRCh38
Transcripts
NM_001142505.1 NM_022169.4 XM_011542952.2 XM_017018121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3868554 c.647T>G p.V216G Chr.11: 119154876-119154876 on Build GRCh38
1492441 c.558G>A p.T186T Chr.11: 119154787-119154787 on Build GRCh38
5526096 c.615G>A p.Q205Q Chr.11: 119154844-119154844 on Build GRCh38
3868553 c.579C>T p.C193C Chr.11: 119154808-119154808 on Build GRCh38
5371802 c.617G>A p.R206K Chr.11: 119154846-119154846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749752428 Chr.11: 119154796-119154796 on Build GRCh38
rs779514029 Chr.11: 119154805-119154805 on Build GRCh38
rs145213898 Chr.11: 119154886-119154886 on Build GRCh38
rs774204201 Chr.11: 119154910-119154910 on Build GRCh38
rs774500849 Chr.11: 119154775-119154775 on Build GRCh38
rs760832720 Chr.11: 119154744-119154744 on Build GRCh38
rs779274927 Chr.11: 119154865-119154865 on Build GRCh38
rs759480287 Chr.11: 119154927-119154927 on Build GRCh38
rs771802760 Chr.11: 119154928-119154928 on Build GRCh38
rs534539509 Chr.11: 119154832-119154832 on Build GRCh38
rs373868091 Chr.11: 119154770-119154770 on Build GRCh38
rs140298630 Chr.11: 119154862-119154862 on Build GRCh38
rs758779632 Chr.11: 119154870-119154870 on Build GRCh38
rs780589874 Chr.11: 119154885-119154885 on Build GRCh38
rs781139493 Chr.11: 119154836-119154836 on Build GRCh38
rs151176056 Chr.11: 119154816-119154816 on Build GRCh38
rs755741606 Chr.11: 119154859-119154859 on Build GRCh38
rs774335112 Chr.11: 119154839-119154839 on Build GRCh38
rs768734717 Chr.11: 119154819-119154819 on Build GRCh38
rs771266308 Chr.11: 119154721-119154721 on Build GRCh38
rs751328446 Chr.11: 119154779-119154779 on Build GRCh38
rs201318219 Chr.11: 119154852-119154852 on Build GRCh38
rs375767553 Chr.11: 119154787-119154787 on Build GRCh38
rs768732954 Chr.11: 119154890-119154890 on Build GRCh38
rs754820331 Chr.11: 119154782-119154782 on Build GRCh38
rs552901604 Chr.11: 119154838-119154838 on Build GRCh38
rs575247461 Chr.11: 119154863-119154863 on Build GRCh38
rs112382702 Chr.11: 119154716-119154716 on Build GRCh38
rs776608146 Chr.11: 119154831-119154831 on Build GRCh38
rs1056041097 Chr.11: 119154712-119154712 on Build GRCh38
rs769978545 Chr.11: 119154908-119154908 on Build GRCh38
rs759467898 Chr.11: 119154786-119154786 on Build GRCh38
rs1005717283 Chr.11: 119154778-119154778 on Build GRCh38
rs141832911 Chr.11: 119154804-119154804 on Build GRCh38
rs776965085 Chr.11: 119154892-119154892 on Build GRCh38
rs772632416 Chr.11: 119154812-119154812 on Build GRCh38
rs149095826 Chr.11: 119154784-119154784 on Build GRCh38
rs569267107 Chr.11: 119154737-119154737 on Build GRCh38
rs11822683 Chr.11: 119154733-119154733 on Build GRCh38
rs775806306 Chr.11: 119154841-119154841 on Build GRCh38
rs774599353 Chr.11: 119154724-119154724 on Build GRCh38
rs752285981 Chr.11: 119154857-119154857 on Build GRCh38
rs754620704 Chr.11: 119154827-119154828 on Build GRCh38
rs370020367 Chr.11: 119154751-119154751 on Build GRCh38
rs200081211 Chr.11: 119154850-119154850 on Build GRCh38
rs147253441 Chr.11: 119154817-119154817 on Build GRCh38
rs368581778 Chr.11: 119154736-119154736 on Build GRCh38
rs749243610 Chr.11: 119154715-119154715 on Build GRCh38

View Full Product Details