Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154821211-154821478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAXDC2
Entrez Gene ID
10826
Gene Name
fatty acid hydroxylase domain containing 2
Gene Aliases
C5orf4
UniGene
Hs.519694
Gene Chromosome Location
Chr.5: 154817018-154850685 on Build GRCh38
Transcripts
NM_032385.4 XM_005268358.2 XM_006714753.2 XM_017008965.1 XM_017008966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6661450 c.728A>G p.H243R Chr.5: 154821377-154821377 on Build GRCh38
1137289 c.747G>A p.M249I Chr.5: 154821358-154821358 on Build GRCh38
7201578 c.733T>C p.S245P Chr.5: 154821372-154821372 on Build GRCh38
1064752 c.812C>A p.P271H Chr.5: 154821293-154821293 on Build GRCh38
5486460 c.829G>A p.D277N Chr.5: 154821276-154821276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758984593 Chr.5: 154821355-154821355 on Build GRCh38
rs779006214 Chr.5: 154821311-154821311 on Build GRCh38
rs748110873 Chr.5: 154821322-154821322 on Build GRCh38
rs762126128 Chr.5: 154821398-154821398 on Build GRCh38
rs1014864317 Chr.5: 154821277-154821277 on Build GRCh38
rs773964447 Chr.5: 154821257-154821257 on Build GRCh38
rs770270682 Chr.5: 154821251-154821251 on Build GRCh38
rs754118338 Chr.5: 154821301-154821301 on Build GRCh38
rs780995046 Chr.5: 154821320-154821320 on Build GRCh38
rs369631535 Chr.5: 154821268-154821268 on Build GRCh38
rs553528180 Chr.5: 154821343-154821343 on Build GRCh38
rs752622206 Chr.5: 154821421-154821421 on Build GRCh38
rs377392941 Chr.5: 154821409-154821409 on Build GRCh38
rs760954923 Chr.5: 154821294-154821294 on Build GRCh38
rs771546322 Chr.5: 154821361-154821361 on Build GRCh38
rs756084643 Chr.5: 154821427-154821427 on Build GRCh38
rs761234799 Chr.5: 154821382-154821382 on Build GRCh38
rs748985025 Chr.5: 154821337-154821337 on Build GRCh38
rs750502118 Chr.5: 154821314-154821314 on Build GRCh38
rs767934081 Chr.5: 154821273-154821273 on Build GRCh38
rs564950368 Chr.5: 154821401-154821401 on Build GRCh38
rs1045879349 Chr.5: 154821289-154821289 on Build GRCh38
rs749114950 Chr.5: 154821453-154821453 on Build GRCh38
rs572907325 Chr.5: 154821262-154821262 on Build GRCh38
rs757448924 Chr.5: 154821303-154821303 on Build GRCh38
rs376650105 Chr.5: 154821325-154821325 on Build GRCh38
rs773064516 Chr.5: 154821237-154821237 on Build GRCh38
rs35592087 Chr.5: 154821308-154821309 on Build GRCh38
rs758472970 Chr.5: 154821411-154821411 on Build GRCh38
rs779697775 Chr.5: 154821405-154821406 on Build GRCh38
rs750591002 Chr.5: 154821410-154821410 on Build GRCh38
rs368827829 Chr.5: 154821412-154821412 on Build GRCh38
rs770735326 Chr.5: 154821345-154821345 on Build GRCh38
rs745408356 Chr.5: 154821331-154821331 on Build GRCh38
rs374973207 Chr.5: 154821239-154821239 on Build GRCh38
rs758444549 Chr.5: 154821319-154821319 on Build GRCh38
rs777810814 Chr.5: 154821432-154821432 on Build GRCh38
rs374577433 Chr.5: 154821376-154821376 on Build GRCh38
rs930716162 Chr.5: 154821244-154821244 on Build GRCh38
rs777684429 Chr.5: 154821327-154821327 on Build GRCh38
rs764441888 Chr.5: 154821298-154821298 on Build GRCh38
rs773906288 Chr.5: 154821295-154821295 on Build GRCh38
rs755574899 Chr.5: 154821351-154821351 on Build GRCh38
rs748785319 Chr.5: 154821335-154821335 on Build GRCh38
rs878965361 Chr.5: 154821310-154821310 on Build GRCh38
rs199687051 Chr.5: 154821346-154821346 on Build GRCh38
rs957074353 Chr.5: 154821356-154821356 on Build GRCh38
rs754171605 Chr.5: 154821396-154821396 on Build GRCh38
rs200568911 Chr.5: 154821276-154821276 on Build GRCh38
rs747826037 Chr.5: 154821386-154821386 on Build GRCh38
rs778318350 Chr.5: 154821266-154821268 on Build GRCh38
rs764491000 Chr.5: 154821393-154821393 on Build GRCh38

View Full Product Details