Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 189984521-189984782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P3H2
Entrez Gene ID
55214
Gene Name
prolyl 3-hydroxylase 2
Gene Aliases
LEPREL1, MCVD, MLAT4
UniGene
Hs.374191
Gene Chromosome Location
Chr.3: 189956728-190122437 on Build GRCh38
Transcripts
NM_001134418.1 NM_018192.3 XM_011512955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1213274 c.1216C>T p.Q406* Chr.3: 189984563-189984563 on Build GRCh38
1042507 c.1228C>T p.R410W Chr.3: 189984551-189984551 on Build GRCh38
1581139 c.1219G>A p.D407N Chr.3: 189984560-189984560 on Build GRCh38
7374929 c.1224G>C p.E408D Chr.3: 189984555-189984555 on Build GRCh38
7111052 c.1222G>C p.E408Q Chr.3: 189984557-189984557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769357547 Chr.3: 189984598-189984598 on Build GRCh38
rs1035225264 Chr.3: 189984709-189984709 on Build GRCh38
rs200097659 Chr.3: 189984615-189984615 on Build GRCh38
rs772552676 Chr.3: 189984573-189984573 on Build GRCh38
rs780583614 Chr.3: 189984577-189984577 on Build GRCh38
rs560125839 Chr.3: 189984608-189984609 on Build GRCh38
rs556193774 Chr.3: 189984655-189984655 on Build GRCh38
rs968003194 Chr.3: 189984750-189984750 on Build GRCh38
rs774914899 Chr.3: 189984550-189984550 on Build GRCh38
rs570298526 Chr.3: 189984586-189984586 on Build GRCh38
rs746578924 Chr.3: 189984583-189984583 on Build GRCh38
rs760758403 Chr.3: 189984608-189984608 on Build GRCh38
rs768389240 Chr.3: 189984584-189984584 on Build GRCh38
rs373492577 Chr.3: 189984603-189984603 on Build GRCh38
rs377600857 Chr.3: 189984566-189984566 on Build GRCh38
rs866386796 Chr.3: 189984634-189984634 on Build GRCh38
rs913932474 Chr.3: 189984747-189984747 on Build GRCh38
rs11311050 Chr.3: 189984607-189984607 on Build GRCh38
rs182815878 Chr.3: 189984682-189984682 on Build GRCh38
rs886762043 Chr.3: 189984675-189984675 on Build GRCh38
rs543887504 Chr.3: 189984607-189984608 on Build GRCh38
rs751385780 Chr.3: 189984606-189984606 on Build GRCh38
rs797004634 Chr.3: 189984617-189984617 on Build GRCh38
rs1046799722 Chr.3: 189984636-189984636 on Build GRCh38
rs988176718 Chr.3: 189984728-189984728 on Build GRCh38
rs547125198 Chr.3: 189984677-189984677 on Build GRCh38
rs746444503 Chr.3: 189984570-189984570 on Build GRCh38
rs577514238 Chr.3: 189984608-189984608 on Build GRCh38
rs558862180 Chr.3: 189984729-189984729 on Build GRCh38
rs1042906768 Chr.3: 189984665-189984665 on Build GRCh38
rs1021927830 Chr.3: 189984614-189984614 on Build GRCh38
rs7610594 Chr.3: 189984693-189984693 on Build GRCh38
rs78427517 Chr.3: 189984616-189984617 on Build GRCh38
rs1016881582 Chr.3: 189984685-189984685 on Build GRCh38
rs766095350 Chr.3: 189984605-189984605 on Build GRCh38
rs768150346 Chr.3: 189984606-189984607 on Build GRCh38
rs377458431 Chr.3: 189984756-189984756 on Build GRCh38
rs757354354 Chr.3: 189984621-189984621 on Build GRCh38
rs35737596 Chr.3: 189984557-189984557 on Build GRCh38
rs374590301 Chr.3: 189984725-189984725 on Build GRCh38
rs1002049526 Chr.3: 189984696-189984696 on Build GRCh38
rs141462471 Chr.3: 189984585-189984585 on Build GRCh38
rs756359436 Chr.3: 189984549-189984549 on Build GRCh38
rs369190383 Chr.3: 189984597-189984597 on Build GRCh38
rs78521896 Chr.3: 189984617-189984617 on Build GRCh38
rs757331420 Chr.3: 189984565-189984565 on Build GRCh38
rs773116869 Chr.3: 189984602-189984602 on Build GRCh38

View Full Product Details