Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1597639-1597912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP5-2
Entrez Gene ID
440021
Gene Name
keratin associated protein 5-2
Gene Aliases
KRTAP5-8, KRTAP5.2
UniGene
Hs.633229
Gene Chromosome Location
Chr.11: 1597177-1598294 on Build GRCh38
Transcripts
NM_001004325.1 NR_021489.1

Gene Symbol
KRTAP5-AS1
Entrez Gene ID
338651
Gene Name
KRTAP5-1/KRTAP5-2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.633229
Gene Chromosome Location
Chr.11: 1572741-1599184 on Build GRCh38
Transcripts
NM_001004325.1 NR_021489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
687102 c.353G>C p.C118S Chr.11: 1597898-1597898 on Build GRCh38
4994951 c.392C>A p.S131Y Chr.11: 1597859-1597859 on Build GRCh38
6698538 c.369C>T p.C123C Chr.11: 1597882-1597882 on Build GRCh38
5838241 c.404C>T p.S135F Chr.11: 1597847-1597847 on Build GRCh38
343192 c.354T>C p.C118C Chr.11: 1597897-1597897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200247586 Chr.11: 1597700-1597700 on Build GRCh38
rs769507671 Chr.11: 1597820-1597820 on Build GRCh38
rs200546709 Chr.11: 1597754-1597754 on Build GRCh38
rs763643856 Chr.11: 1597746-1597746 on Build GRCh38
rs747078036 Chr.11: 1597804-1597804 on Build GRCh38
rs764015892 Chr.11: 1597853-1597853 on Build GRCh38
rs751056617 Chr.11: 1597748-1597748 on Build GRCh38
rs376517981 Chr.11: 1597884-1597884 on Build GRCh38
rs201524140 Chr.11: 1597741-1597741 on Build GRCh38
rs769620553 Chr.11: 1597808-1597837 on Build GRCh38
rs748619375 Chr.11: 1597699-1597699 on Build GRCh38
rs1019747468 Chr.11: 1597846-1597846 on Build GRCh38
rs750036276 Chr.11: 1597770-1597770 on Build GRCh38
rs267602793 Chr.11: 1597701-1597701 on Build GRCh38
rs878891440 Chr.11: 1597828-1597828 on Build GRCh38
rs549098367 Chr.11: 1597817-1597817 on Build GRCh38
rs541700992 Chr.11: 1597755-1597755 on Build GRCh38
rs756308920 Chr.11: 1597680-1597680 on Build GRCh38
rs201023715 Chr.11: 1597768-1597768 on Build GRCh38
rs778053349 Chr.11: 1597801-1597801 on Build GRCh38
rs371816197 Chr.11: 1597843-1597843 on Build GRCh38
rs755324312 Chr.11: 1597778-1597778 on Build GRCh38
rs544007188 Chr.11: 1597670-1597670 on Build GRCh38
rs775296238 Chr.11: 1597844-1597845 on Build GRCh38
rs776553426 Chr.11: 1597726-1597726 on Build GRCh38
rs765022489 Chr.11: 1597735-1597735 on Build GRCh38
rs780527293 Chr.11: 1597667-1597667 on Build GRCh38
rs370177579 Chr.11: 1597888-1597888 on Build GRCh38
rs777215305 Chr.11: 1597658-1597658 on Build GRCh38
rs750071358 Chr.11: 1597870-1597870 on Build GRCh38
rs774208637 Chr.11: 1597857-1597857 on Build GRCh38
rs138473551 Chr.11: 1597842-1597842 on Build GRCh38
rs762836364 Chr.11: 1597743-1597743 on Build GRCh38
rs761241217 Chr.11: 1597860-1597860 on Build GRCh38
rs151223920 Chr.11: 1597862-1597862 on Build GRCh38
rs527259468 Chr.11: 1597806-1597806 on Build GRCh38
rs145895339 Chr.11: 1597662-1597662 on Build GRCh38
rs779300507 Chr.11: 1597797-1597797 on Build GRCh38
rs771042603 Chr.11: 1597723-1597723 on Build GRCh38
rs747280175 Chr.11: 1597715-1597715 on Build GRCh38
rs565622148 Chr.11: 1597673-1597673 on Build GRCh38
rs141027563 Chr.11: 1597800-1597800 on Build GRCh38
rs773399887 Chr.11: 1597706-1597706 on Build GRCh38
rs756881125 Chr.11: 1597664-1597664 on Build GRCh38
rs374220501 Chr.11: 1597847-1597847 on Build GRCh38
rs560118269 Chr.11: 1597805-1597805 on Build GRCh38
rs779303748 Chr.11: 1597690-1597690 on Build GRCh38
rs775311215 Chr.11: 1597738-1597738 on Build GRCh38
rs746052009 Chr.11: 1597819-1597819 on Build GRCh38
rs199659307 Chr.11: 1597750-1597750 on Build GRCh38

View Full Product Details