Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 122213425-122213670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHX6
Entrez Gene ID
26468
Gene Name
LIM homeobox 6
Gene Aliases
LHX6.1
UniGene
Hs.103137
Gene Chromosome Location
Chr.9: 122185626-122228893 on Build GRCh38
Transcripts
NM_001242333.1 NM_001242334.1 NM_001242335.1 NM_014368.4 NM_199160.3 XM_011518521.2 XM_011518522.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402093 c.1008C>G p.S336R Chr.9: 122213652-122213652 on Build GRCh38
4974478 c.1042G>A p.G348S Chr.9: 122213618-122213618 on Build GRCh38
4877405 c.1046A>G p.Y349C Chr.9: 122213614-122213614 on Build GRCh38
4974477 c.1042G>A p.G348S Chr.9: 122213618-122213618 on Build GRCh38
6402092 c.1008C>G p.S336R Chr.9: 122213652-122213652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750456770 Chr.9: 122213587-122213587 on Build GRCh38
rs1029678696 Chr.9: 122213534-122213534 on Build GRCh38
rs779843806 Chr.9: 122213615-122213615 on Build GRCh38
rs143968888 Chr.9: 122213509-122213509 on Build GRCh38
rs753014963 Chr.9: 122213558-122213558 on Build GRCh38
rs747634996 Chr.9: 122213646-122213646 on Build GRCh38
rs966552638 Chr.9: 122213476-122213476 on Build GRCh38
rs780731428 Chr.9: 122213633-122213633 on Build GRCh38
rs746787054 Chr.9: 122213616-122213616 on Build GRCh38
rs375958601 Chr.9: 122213632-122213632 on Build GRCh38
rs547254073 Chr.9: 122213497-122213497 on Build GRCh38
rs895021061 Chr.9: 122213451-122213451 on Build GRCh38
rs529487663 Chr.9: 122213510-122213510 on Build GRCh38
rs767926264 Chr.9: 122213556-122213556 on Build GRCh38
rs999588618 Chr.9: 122213523-122213523 on Build GRCh38
rs1032192935 Chr.9: 122213491-122213491 on Build GRCh38
rs550936469 Chr.9: 122213567-122213567 on Build GRCh38
rs974724311 Chr.9: 122213588-122213588 on Build GRCh38
rs569120489 Chr.9: 122213570-122213570 on Build GRCh38
rs758258901 Chr.9: 122213597-122213597 on Build GRCh38
rs1018946172 Chr.9: 122213566-122213566 on Build GRCh38
rs780118827 Chr.9: 122213478-122213478 on Build GRCh38
rs200272141 Chr.9: 122213580-122213580 on Build GRCh38
rs764251235 Chr.9: 122213569-122213569 on Build GRCh38
rs955551720 Chr.9: 122213540-122213540 on Build GRCh38
rs1021152805 Chr.9: 122213459-122213459 on Build GRCh38
rs769359783 Chr.9: 122213648-122213648 on Build GRCh38
rs528508332 Chr.9: 122213488-122213488 on Build GRCh38

View Full Product Details