Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133186586-133186811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KNDC1
Entrez Gene ID
85442
Gene Name
kinase non-catalytic C-lobe domain containing 1
Gene Aliases
C10orf23, RASGEF2, bB439H18.3
UniGene
Hs.530685
Gene Chromosome Location
Chr.10: 133160096-133226412 on Build GRCh38
Transcripts
NM_152643.6 XM_017016858.1 XM_017016859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4175725 c.1272C>T p.D424D Chr.10: 133186620-133186620 on Build GRCh38
4175726 c.1304T>A p.L435Q Chr.10: 133186652-133186652 on Build GRCh38
6697518 c.1320G>A p.A440A Chr.10: 133186668-133186668 on Build GRCh38
6128590 c.1258C>T p.Q420* Chr.10: 133186606-133186606 on Build GRCh38
3751653 c.1307A>G p.E436G Chr.10: 133186655-133186655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773005915 Chr.10: 133186654-133186654 on Build GRCh38
rs943644623 Chr.10: 133186686-133186686 on Build GRCh38
rs764143392 Chr.10: 133186726-133186726 on Build GRCh38
rs771982127 Chr.10: 133186646-133186646 on Build GRCh38
rs549726304 Chr.10: 133186711-133186711 on Build GRCh38
rs750762105 Chr.10: 133186681-133186681 on Build GRCh38
rs768280562 Chr.10: 133186705-133186705 on Build GRCh38
rs1021277868 Chr.10: 133186635-133186635 on Build GRCh38
rs781755735 Chr.10: 133186614-133186614 on Build GRCh38
rs1053069592 Chr.10: 133186737-133186737 on Build GRCh38
rs369446107 Chr.10: 133186708-133186708 on Build GRCh38
rs757586250 Chr.10: 133186611-133186611 on Build GRCh38
rs766297714 Chr.10: 133186665-133186665 on Build GRCh38
rs780588892 Chr.10: 133186684-133186684 on Build GRCh38
rs3810964 Chr.10: 133186655-133186655 on Build GRCh38
rs1046701447 Chr.10: 133186727-133186727 on Build GRCh38
rs764611825 Chr.10: 133186769-133186769 on Build GRCh38
rs151145298 Chr.10: 133186755-133186755 on Build GRCh38
rs763086309 Chr.10: 133186715-133186715 on Build GRCh38
rs541977090 Chr.10: 133186630-133186630 on Build GRCh38
rs778768611 Chr.10: 133186706-133186706 on Build GRCh38
rs372254199 Chr.10: 133186666-133186666 on Build GRCh38
rs563461634 Chr.10: 133186713-133186713 on Build GRCh38
rs764339506 Chr.10: 133186667-133186667 on Build GRCh38
rs774119251 Chr.10: 133186631-133186631 on Build GRCh38
rs563157577 Chr.10: 133186714-133186714 on Build GRCh38
rs376829471 Chr.10: 133186692-133186692 on Build GRCh38
rs748758379 Chr.10: 133186627-133186627 on Build GRCh38
rs367788279 Chr.10: 133186620-133186620 on Build GRCh38
rs1020652828 Chr.10: 133186733-133186733 on Build GRCh38
rs775049092 Chr.10: 133186710-133186710 on Build GRCh38
rs746345911 Chr.10: 133186617-133186617 on Build GRCh38
rs752928995 Chr.10: 133186732-133186732 on Build GRCh38
rs760794428 Chr.10: 133186754-133186754 on Build GRCh38
rs3810965 Chr.10: 133186644-133186644 on Build GRCh38
rs757674308 Chr.10: 133186668-133186668 on Build GRCh38
rs199618846 Chr.10: 133186675-133186675 on Build GRCh38
rs935523667 Chr.10: 133186736-133186736 on Build GRCh38
rs891424768 Chr.10: 133186621-133186621 on Build GRCh38
rs954556956 Chr.10: 133186671-133186671 on Build GRCh38

View Full Product Details