Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120399782-120400055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL1
Entrez Gene ID
11167
Gene Name
follistatin like 1
Gene Aliases
FRP, FSL1, MIR198, OCC-1, OCC1, tsc36
UniGene
Hs.269512
Gene Chromosome Location
Chr.3: 120394214-120451071 on Build GRCh38
Transcripts
NM_007085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5559205 c.869T>A p.L290H Chr.3: 120399896-120399896 on Build GRCh38
6433239 c.862C>A p.Q288K Chr.3: 120399903-120399903 on Build GRCh38
1418084 c.861C>A p.V287V Chr.3: 120399904-120399904 on Build GRCh38
7368903 c.865G>C p.E289Q Chr.3: 120399900-120399900 on Build GRCh38
1036855 c.832C>A p.Q278K Chr.3: 120399933-120399933 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780033290 Chr.3: 120399886-120399886 on Build GRCh38
rs748295626 Chr.3: 120399887-120399887 on Build GRCh38
rs761360155 Chr.3: 120399987-120399987 on Build GRCh38
rs919636749 Chr.3: 120399900-120399900 on Build GRCh38
rs755932324 Chr.3: 120399884-120399884 on Build GRCh38
rs767168158 Chr.3: 120399988-120399988 on Build GRCh38
rs773156885 Chr.3: 120399910-120399910 on Build GRCh38
rs553209646 Chr.3: 120399833-120399833 on Build GRCh38
rs762073333 Chr.3: 120399840-120399840 on Build GRCh38
rs775164475 Chr.3: 120399929-120399929 on Build GRCh38
rs751186231 Chr.3: 120399961-120399961 on Build GRCh38
rs996309220 Chr.3: 120400027-120400027 on Build GRCh38
rs145080646 Chr.3: 120399907-120399907 on Build GRCh38
rs989459231 Chr.3: 120399929-120399929 on Build GRCh38
rs573152399 Chr.3: 120399905-120399905 on Build GRCh38
rs779980432 Chr.3: 120400009-120400009 on Build GRCh38
rs573608027 Chr.3: 120399866-120399866 on Build GRCh38
rs750352523 Chr.3: 120400004-120400004 on Build GRCh38
rs747040181 Chr.3: 120399911-120399911 on Build GRCh38
rs375335990 Chr.3: 120399879-120399879 on Build GRCh38
rs763735659 Chr.3: 120399955-120399955 on Build GRCh38
rs760079187 Chr.3: 120399946-120399946 on Build GRCh38
rs756725039 Chr.3: 120399872-120399872 on Build GRCh38
rs141248889 Chr.3: 120399883-120399883 on Build GRCh38
rs541862974 Chr.3: 120399949-120399949 on Build GRCh38
rs370317312 Chr.3: 120399843-120399843 on Build GRCh38
rs751149012 Chr.3: 120399870-120399870 on Build GRCh38
rs368394559 Chr.3: 120400018-120400018 on Build GRCh38
rs564819042 Chr.3: 120400017-120400017 on Build GRCh38
rs373021638 Chr.3: 120399928-120399928 on Build GRCh38
rs772403305 Chr.3: 120399926-120399928 on Build GRCh38
rs771009480 Chr.3: 120399914-120399914 on Build GRCh38
rs147284645 Chr.3: 120399822-120399822 on Build GRCh38
rs963572775 Chr.3: 120399820-120399820 on Build GRCh38
rs528267070 Chr.3: 120400014-120400014 on Build GRCh38
rs377558847 Chr.3: 120399972-120399972 on Build GRCh38
rs371812718 Chr.3: 120399951-120399951 on Build GRCh38
rs550670030 Chr.3: 120400023-120400023 on Build GRCh38
rs544928601 Chr.3: 120400015-120400015 on Build GRCh38
rs569290520 Chr.3: 120400006-120400006 on Build GRCh38
rs759833891 Chr.3: 120399835-120399835 on Build GRCh38
rs765624624 Chr.3: 120399837-120399837 on Build GRCh38
rs781219887 Chr.3: 120399873-120399873 on Build GRCh38
rs915259827 Chr.3: 120399934-120399934 on Build GRCh38
rs542815805 Chr.3: 120399867-120399867 on Build GRCh38

View Full Product Details