Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66859473-66859712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRFN4
Entrez Gene ID
78999
Gene Name
leucine rich repeat and fibronectin type III domain containing 4
Gene Aliases
FIGLER6, SALM3, SALM3.
UniGene
Hs.209979
Gene Chromosome Location
Chr.11: 66857087-66860475 on Build GRCh38
Transcripts
NM_024036.4 XM_005274239.3 NM_000920.3 NM_001040716.1 NM_022172.2 XM_005274031.4 XM_005274032.4 XM_006718578.3 XM_011545086.2 XM_017017868.1 XM_017017869.1 XM_017017870.1 XM_017017871.1 XM_017017872.1

Gene Symbol
PC
Entrez Gene ID
5091
Gene Name
pyruvate carboxylase
Gene Aliases
PCB
UniGene
Hs.89890
Gene Chromosome Location
Chr.11: 66848522-66958418 on Build GRCh38
Transcripts
NM_024036.4 XM_005274239.3 NM_000920.3 NM_001040716.1 NM_022172.2 XM_005274031.4 XM_005274032.4 XM_006718578.3 XM_011545086.2 XM_017017868.1 XM_017017869.1 XM_017017870.1 XM_017017871.1 XM_017017872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027430 c.1378C>T p.L460L Chr.11: 66859665-66859665 on Build GRCh38
5726994 c.1377C>T p.F459F Chr.11: 66859664-66859664 on Build GRCh38
5015029 c.1360G>T p.A454S Chr.11: 66859647-66859647 on Build GRCh38
5970318 c.1359A>C p.P453P Chr.11: 66859646-66859646 on Build GRCh38
5631708 c.1354G>C p.V452L Chr.11: 66859641-66859641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766574016 Chr.11: 66859607-66859607 on Build GRCh38
rs202226738 Chr.11: 66859686-66859686 on Build GRCh38
rs774890962 Chr.11: 66859648-66859648 on Build GRCh38
rs766696740 Chr.11: 66859683-66859683 on Build GRCh38
rs777761696 Chr.11: 66859615-66859615 on Build GRCh38
rs200469470 Chr.11: 66859608-66859608 on Build GRCh38
rs751355859 Chr.11: 66859619-66859619 on Build GRCh38
rs757055957 Chr.11: 66859620-66859620 on Build GRCh38
rs769555912 Chr.11: 66859690-66859693 on Build GRCh38
rs769367704 Chr.11: 66859626-66859626 on Build GRCh38
rs559954859 Chr.11: 66859548-66859548 on Build GRCh38
rs779550031 Chr.11: 66859632-66859632 on Build GRCh38
rs745554440 Chr.11: 66859625-66859625 on Build GRCh38
rs374875298 Chr.11: 66859586-66859586 on Build GRCh38
rs750635863 Chr.11: 66859599-66859599 on Build GRCh38
rs547211574 Chr.11: 66859529-66859529 on Build GRCh38
rs555737259 Chr.11: 66859549-66859549 on Build GRCh38
rs923401798 Chr.11: 66859561-66859561 on Build GRCh38
rs189468338 Chr.11: 66859688-66859688 on Build GRCh38
rs376456123 Chr.11: 66859527-66859527 on Build GRCh38
rs868191334 Chr.11: 66859493-66859493 on Build GRCh38
rs769105380 Chr.11: 66859646-66859646 on Build GRCh38
rs767671905 Chr.11: 66859589-66859589 on Build GRCh38
rs962014971 Chr.11: 66859630-66859630 on Build GRCh38
rs199571830 Chr.11: 66859610-66859610 on Build GRCh38
rs962417135 Chr.11: 66859532-66859532 on Build GRCh38
rs748779643 Chr.11: 66859644-66859644 on Build GRCh38
rs932259122 Chr.11: 66859568-66859568 on Build GRCh38
rs924967361 Chr.11: 66859573-66859573 on Build GRCh38
rs151122985 Chr.11: 66859680-66859680 on Build GRCh38
rs367994202 Chr.11: 66859685-66859685 on Build GRCh38
rs972382275 Chr.11: 66859660-66859660 on Build GRCh38
rs976281386 Chr.11: 66859557-66859557 on Build GRCh38
rs780804776 Chr.11: 66859624-66859624 on Build GRCh38
rs760928802 Chr.11: 66859604-66859604 on Build GRCh38
rs1043974488 Chr.11: 66859496-66859496 on Build GRCh38
rs763991079 Chr.11: 66859692-66859692 on Build GRCh38
rs371094687 Chr.11: 66859679-66859679 on Build GRCh38
rs575841446 Chr.11: 66859569-66859569 on Build GRCh38
rs910272932 Chr.11: 66859687-66859687 on Build GRCh38
rs952029317 Chr.11: 66859510-66859510 on Build GRCh38
rs969226443 Chr.11: 66859577-66859577 on Build GRCh38
rs542518914 Chr.11: 66859544-66859544 on Build GRCh38

View Full Product Details