Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111195835-111196108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND2D
Entrez Gene ID
79961
Gene Name
DENN domain containing 2D
Gene Aliases
-
UniGene
Hs.557850
Gene Chromosome Location
Chr.1: 111185969-111204791 on Build GRCh38
Transcripts
NM_001271833.1 NM_024901.4 XM_006710921.2 XM_006710922.1 XM_006710923.1 XM_006710924.1 XM_011542187.1 XM_011542189.1 XM_011542190.2 XM_017002386.1 XM_017002387.1 XM_017002388.1 XM_017002389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
256152 c.643G>A p.E215K Chr.1: 111195918-111195918 on Build GRCh38
6642337 c.642T>C p.T214T Chr.1: 111195919-111195919 on Build GRCh38
4910744 c.636A>T p.S212S Chr.1: 111195925-111195925 on Build GRCh38
4633250 c.638G>A p.G213D Chr.1: 111195923-111195923 on Build GRCh38
3965697 c.508C>G p.L170V Chr.1: 111196053-111196053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202237373 Chr.1: 111196062-111196062 on Build GRCh38
rs771650458 Chr.1: 111195935-111195935 on Build GRCh38
rs773297086 Chr.1: 111196089-111196089 on Build GRCh38
rs370888665 Chr.1: 111196084-111196084 on Build GRCh38
rs762206224 Chr.1: 111195980-111195980 on Build GRCh38
rs557909053 Chr.1: 111195986-111195986 on Build GRCh38
rs182195264 Chr.1: 111196082-111196082 on Build GRCh38
rs780941523 Chr.1: 111196076-111196076 on Build GRCh38
rs753659411 Chr.1: 111196013-111196013 on Build GRCh38
rs935102065 Chr.1: 111196027-111196027 on Build GRCh38
rs776002368 Chr.1: 111195952-111195952 on Build GRCh38
rs756317104 Chr.1: 111195875-111195875 on Build GRCh38
rs770698608 Chr.1: 111195950-111195950 on Build GRCh38
rs749698610 Chr.1: 111196081-111196081 on Build GRCh38
rs777488040 Chr.1: 111196071-111196071 on Build GRCh38
rs772572581 Chr.1: 111195941-111195941 on Build GRCh38
rs754764614 Chr.1: 111196024-111196024 on Build GRCh38
rs779891645 Chr.1: 111195899-111195899 on Build GRCh38
rs960422460 Chr.1: 111196019-111196019 on Build GRCh38
rs754245478 Chr.1: 111196075-111196075 on Build GRCh38
rs774879110 Chr.1: 111195977-111195977 on Build GRCh38
rs1019026114 Chr.1: 111195857-111195857 on Build GRCh38
rs969493515 Chr.1: 111196050-111196050 on Build GRCh38
rs201335642 Chr.1: 111196085-111196085 on Build GRCh38
rs746714015 Chr.1: 111195944-111195944 on Build GRCh38
rs750694297 Chr.1: 111195874-111195874 on Build GRCh38
rs991933642 Chr.1: 111196031-111196031 on Build GRCh38
rs1017646135 Chr.1: 111195971-111195971 on Build GRCh38
rs34022087 Chr.1: 111196075-111196076 on Build GRCh38
rs756333845 Chr.1: 111195991-111195991 on Build GRCh38
rs1007136725 Chr.1: 111195969-111195969 on Build GRCh38
rs1046174954 Chr.1: 111195992-111195992 on Build GRCh38
rs34558744 Chr.1: 111195967-111195967 on Build GRCh38
rs754925518 Chr.1: 111195929-111195929 on Build GRCh38
rs778538719 Chr.1: 111196036-111196036 on Build GRCh38
rs149018314 Chr.1: 111196003-111196003 on Build GRCh38
rs762729615 Chr.1: 111195954-111195954 on Build GRCh38
rs149711237 Chr.1: 111195931-111195931 on Build GRCh38
rs749020724 Chr.1: 111195910-111195910 on Build GRCh38
rs920618467 Chr.1: 111195942-111195942 on Build GRCh38
rs377337878 Chr.1: 111195961-111195961 on Build GRCh38
rs746452557 Chr.1: 111196072-111196072 on Build GRCh38
rs780768846 Chr.1: 111195863-111195863 on Build GRCh38
rs972441980 Chr.1: 111195934-111195934 on Build GRCh38
rs750498645 Chr.1: 111195990-111195990 on Build GRCh38
rs1023286553 Chr.1: 111196034-111196034 on Build GRCh38
rs150577405 Chr.1: 111195930-111195930 on Build GRCh38

View Full Product Details