Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50746493-50746750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LUC7L3
Entrez Gene ID
51747
Gene Name
LUC7 like 3 pre-mRNA splicing factor
Gene Aliases
CRA, CREAP-1, CROP, LUC7A, OA48-18, hLuc7A
UniGene
Hs.130293
Gene Chromosome Location
Chr.17: 50719565-50754269 on Build GRCh38
Transcripts
NM_006107.3 NM_016424.4 XM_005257448.1 XM_005257449.2 XM_005257452.1 XM_006721943.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6035687 c.759T>A p.D253E Chr.17: 50746551-50746551 on Build GRCh38
706880 c.1095C>T p.S365S Chr.17: 50746659-50746659 on Build GRCh38
6705168 c.1043G>A p.R348Q Chr.17: 50746607-50746607 on Build GRCh38
6705158 c.839G>A p.R280Q Chr.17: 50746631-50746631 on Build GRCh38
1256999 c.994A>G p.R332G Chr.17: 50746558-50746558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559723690 Chr.17: 50746645-50746645 on Build GRCh38
rs774068592 Chr.17: 50746643-50746643 on Build GRCh38
rs750723501 Chr.17: 50746668-50746668 on Build GRCh38
rs781770985 Chr.17: 50746688-50746688 on Build GRCh38
rs754827978 Chr.17: 50746524-50746524 on Build GRCh38
rs373348894 Chr.17: 50746637-50746637 on Build GRCh38
rs111730180 Chr.17: 50746539-50746539 on Build GRCh38
rs774725237 Chr.17: 50746613-50746615 on Build GRCh38
rs778790038 Chr.17: 50746667-50746667 on Build GRCh38
rs532705960 Chr.17: 50746631-50746631 on Build GRCh38
rs779216847 Chr.17: 50746592-50746592 on Build GRCh38
rs749332609 Chr.17: 50746560-50746560 on Build GRCh38
rs757242030 Chr.17: 50746577-50746577 on Build GRCh38
rs747349570 Chr.17: 50746677-50746677 on Build GRCh38
rs773733723 Chr.17: 50746717-50746717 on Build GRCh38
rs934713490 Chr.17: 50746636-50746636 on Build GRCh38
rs759279084 Chr.17: 50746720-50746720 on Build GRCh38
rs746016837 Chr.17: 50746593-50746593 on Build GRCh38
rs367770291 Chr.17: 50746613-50746613 on Build GRCh38
rs767160992 Chr.17: 50746537-50746537 on Build GRCh38
rs970215402 Chr.17: 50746601-50746601 on Build GRCh38
rs916065489 Chr.17: 50746574-50746574 on Build GRCh38
rs770483352 Chr.17: 50746716-50746716 on Build GRCh38
rs112100409 Chr.17: 50746703-50746703 on Build GRCh38
rs1026336013 Chr.17: 50746523-50746523 on Build GRCh38
rs528707738 Chr.17: 50746661-50746661 on Build GRCh38
rs753666290 Chr.17: 50746666-50746666 on Build GRCh38
rs551789475 Chr.17: 50746551-50746551 on Build GRCh38
rs777875119 Chr.17: 50746552-50746552 on Build GRCh38
rs200539321 Chr.17: 50746644-50746644 on Build GRCh38
rs756081157 Chr.17: 50746646-50746646 on Build GRCh38
rs372521753 Chr.17: 50746673-50746673 on Build GRCh38
rs769103676 Chr.17: 50746684-50746684 on Build GRCh38
rs748547330 Chr.17: 50746695-50746695 on Build GRCh38
rs771324379 Chr.17: 50746606-50746629 on Build GRCh38
rs866565901 Chr.17: 50746570-50746570 on Build GRCh38
rs148541531 Chr.17: 50746558-50746558 on Build GRCh38
rs370778632 Chr.17: 50746669-50746669 on Build GRCh38
rs747578523 Chr.17: 50746607-50746607 on Build GRCh38
rs567110880 Chr.17: 50746606-50746606 on Build GRCh38
rs772489923 Chr.17: 50746604-50746604 on Build GRCh38

View Full Product Details