Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35295481-35295750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAG
Entrez Gene ID
4099
Gene Name
myelin associated glycoprotein
Gene Aliases
GMA, S-MAG, SIGLEC-4A, SIGLEC4A, SPG75
UniGene
Hs.643440
Gene Chromosome Location
Chr.19: 35292086-35313807 on Build GRCh38
Transcripts
NM_001199216.1 NM_002361.3 NM_080600.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3892293 c.51C>T p.S17S Chr.19: 35295617-35295617 on Build GRCh38
4497979 c.50C>T p.S17F Chr.19: 35295616-35295616 on Build GRCh38
6859199 c.168T>A p.H56Q Chr.19: 35295734-35295734 on Build GRCh38
1214204 c.178T>C p.Y60H Chr.19: 35295744-35295744 on Build GRCh38
187847 c.157G>A p.A53T Chr.19: 35295723-35295723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200076895 Chr.19: 35295625-35295625 on Build GRCh38
rs201184040 Chr.19: 35295635-35295635 on Build GRCh38
rs1006690864 Chr.19: 35295536-35295536 on Build GRCh38
rs776468293 Chr.19: 35295603-35295603 on Build GRCh38
rs201355497 Chr.19: 35295568-35295568 on Build GRCh38
rs370255945 Chr.19: 35295586-35295586 on Build GRCh38
rs200065531 Chr.19: 35295716-35295716 on Build GRCh38
rs201620798 Chr.19: 35295729-35295729 on Build GRCh38
rs137856043 Chr.19: 35295619-35295619 on Build GRCh38
rs568741601 Chr.19: 35295549-35295549 on Build GRCh38
rs758882531 Chr.19: 35295587-35295587 on Build GRCh38
rs200578769 Chr.19: 35295567-35295567 on Build GRCh38
rs11550211 Chr.19: 35295662-35295662 on Build GRCh38
rs879138750 Chr.19: 35295539-35295539 on Build GRCh38
rs765732010 Chr.19: 35295695-35295695 on Build GRCh38
rs754048834 Chr.19: 35295579-35295579 on Build GRCh38
rs778004595 Chr.19: 35295588-35295588 on Build GRCh38
rs199932400 Chr.19: 35295608-35295608 on Build GRCh38
rs768376515 Chr.19: 35295576-35295577 on Build GRCh38
rs1018543313 Chr.19: 35295544-35295544 on Build GRCh38
rs540917777 Chr.19: 35295677-35295677 on Build GRCh38
rs200281744 Chr.19: 35295694-35295694 on Build GRCh38
rs200718920 Chr.19: 35295718-35295718 on Build GRCh38
rs200850404 Chr.19: 35295594-35295594 on Build GRCh38
rs779021354 Chr.19: 35295583-35295583 on Build GRCh38
rs201907391 Chr.19: 35295711-35295711 on Build GRCh38
rs201932537 Chr.19: 35295611-35295611 on Build GRCh38
rs200461020 Chr.19: 35295714-35295714 on Build GRCh38
rs759758253 Chr.19: 35295723-35295723 on Build GRCh38
rs147119099 Chr.19: 35295609-35295609 on Build GRCh38
rs768040433 Chr.19: 35295709-35295709 on Build GRCh38
rs755101115 Chr.19: 35295581-35295581 on Build GRCh38
rs925330167 Chr.19: 35295572-35295572 on Build GRCh38
rs199802808 Chr.19: 35295570-35295570 on Build GRCh38
rs751314676 Chr.19: 35295562-35295562 on Build GRCh38
rs201798784 Chr.19: 35295682-35295682 on Build GRCh38
rs918875166 Chr.19: 35295634-35295634 on Build GRCh38
rs965295757 Chr.19: 35295545-35295545 on Build GRCh38
rs993871861 Chr.19: 35295520-35295520 on Build GRCh38
rs986825704 Chr.19: 35295659-35295659 on Build GRCh38
rs761655490 Chr.19: 35295624-35295624 on Build GRCh38
rs754100178 Chr.19: 35295636-35295636 on Build GRCh38
rs370825816 Chr.19: 35295627-35295627 on Build GRCh38
rs201203478 Chr.19: 35295674-35295674 on Build GRCh38
rs998499675 Chr.19: 35295552-35295552 on Build GRCh38
rs888206774 Chr.19: 35295530-35295530 on Build GRCh38
rs764028764 Chr.19: 35295614-35295614 on Build GRCh38
rs757733728 Chr.19: 35295678-35295678 on Build GRCh38
rs745482627 Chr.19: 35295606-35295606 on Build GRCh38
rs1048163692 Chr.19: 35295521-35295521 on Build GRCh38
rs373592019 Chr.19: 35295511-35295511 on Build GRCh38
rs765244771 Chr.19: 35295658-35295658 on Build GRCh38
rs745533880 Chr.19: 35295693-35295693 on Build GRCh38
rs761708648 Chr.19: 35295717-35295717 on Build GRCh38
rs201061871 Chr.19: 35295701-35295701 on Build GRCh38
rs752755510 Chr.19: 35295666-35295666 on Build GRCh38
rs374238503 Chr.19: 35295686-35295686 on Build GRCh38
rs201985530 Chr.19: 35295596-35295596 on Build GRCh38
rs929595537 Chr.19: 35295654-35295654 on Build GRCh38
rs202017359 Chr.19: 35295650-35295650 on Build GRCh38
rs966680288 Chr.19: 35295555-35295555 on Build GRCh38
rs552073048 Chr.19: 35295535-35295535 on Build GRCh38
rs537546335 Chr.19: 35295554-35295554 on Build GRCh38
rs772943178 Chr.19: 35295722-35295722 on Build GRCh38
rs143777259 Chr.19: 35295671-35295671 on Build GRCh38

View Full Product Details