Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 165544475-165544730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC400794
Entrez Gene ID
400794
Gene Name
uncharacterized LOC400794
Gene Aliases
-
UniGene
Hs.374847
Gene Chromosome Location
Chr.1: 165476842-165582155 on Build GRCh38
Transcripts
NR_026744.2 NM_001005214.3

Gene Symbol
LRRC52
Entrez Gene ID
440699
Gene Name
leucine rich repeat containing 52
Gene Aliases
-
UniGene
Hs.664975
Gene Chromosome Location
Chr.1: 165544241-165563948 on Build GRCh38
Transcripts
NR_026744.2 NM_001005214.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3710390 c.217C>G p.L73V Chr.1: 165544513-165544513 on Build GRCh38
145598 c.308T>A p.L103H Chr.1: 165544604-165544604 on Build GRCh38
463330 c.417A>T p.L139F Chr.1: 165544713-165544713 on Build GRCh38
6703977 c.327C>T p.S109S Chr.1: 165544623-165544623 on Build GRCh38
1499432 c.363C>T p.F121F Chr.1: 165544659-165544659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373153274 Chr.1: 165544656-165544656 on Build GRCh38
rs140229260 Chr.1: 165544678-165544678 on Build GRCh38
rs761897498 Chr.1: 165544702-165544702 on Build GRCh38
rs778453459 Chr.1: 165544548-165544554 on Build GRCh38
rs768507595 Chr.1: 165544651-165544651 on Build GRCh38
rs753065284 Chr.1: 165544594-165544594 on Build GRCh38
rs756599761 Chr.1: 165544610-165544610 on Build GRCh38
rs745990615 Chr.1: 165544632-165544632 on Build GRCh38
rs898106121 Chr.1: 165544605-165544605 on Build GRCh38
rs758219032 Chr.1: 165544637-165544637 on Build GRCh38
rs775106406 Chr.1: 165544558-165544558 on Build GRCh38
rs764714371 Chr.1: 165544567-165544567 on Build GRCh38
rs754255011 Chr.1: 165544699-165544699 on Build GRCh38
rs143080110 Chr.1: 165544662-165544662 on Build GRCh38
rs140364875 Chr.1: 165544612-165544612 on Build GRCh38
rs749360058 Chr.1: 165544506-165544506 on Build GRCh38
rs367635989 Chr.1: 165544559-165544559 on Build GRCh38
rs922303973 Chr.1: 165544682-165544682 on Build GRCh38
rs758342059 Chr.1: 165544661-165544662 on Build GRCh38
rs1025568623 Chr.1: 165544616-165544616 on Build GRCh38
rs533989036 Chr.1: 165544629-165544629 on Build GRCh38
rs114878977 Chr.1: 165544564-165544564 on Build GRCh38
rs376333861 Chr.1: 165544663-165544663 on Build GRCh38
rs1050020915 Chr.1: 165544529-165544529 on Build GRCh38
rs149047999 Chr.1: 165544641-165544641 on Build GRCh38
rs764403645 Chr.1: 165544611-165544611 on Build GRCh38
rs745730834 Chr.1: 165544522-165544522 on Build GRCh38
rs750392324 Chr.1: 165544646-165544646 on Build GRCh38
rs115653001 Chr.1: 165544542-165544542 on Build GRCh38
rs753418467 Chr.1: 165544521-165544521 on Build GRCh38
rs144317602 Chr.1: 165544695-165544695 on Build GRCh38
rs375094839 Chr.1: 165544521-165544521 on Build GRCh38
rs906623372 Chr.1: 165544596-165544596 on Build GRCh38
rs951031179 Chr.1: 165544688-165544688 on Build GRCh38
rs201605916 Chr.1: 165544661-165544661 on Build GRCh38
rs114654661 Chr.1: 165544593-165544593 on Build GRCh38
rs376324521 Chr.1: 165544505-165544505 on Build GRCh38
rs936585684 Chr.1: 165544534-165544534 on Build GRCh38
rs369144835 Chr.1: 165544590-165544590 on Build GRCh38
rs757475818 Chr.1: 165544517-165544517 on Build GRCh38
rs758472016 Chr.1: 165544668-165544668 on Build GRCh38
rs370972088 Chr.1: 165544592-165544592 on Build GRCh38
rs374982726 Chr.1: 165544565-165544565 on Build GRCh38
rs757562327 Chr.1: 165544623-165544623 on Build GRCh38
rs1011175465 Chr.1: 165544660-165544660 on Build GRCh38
rs756873698 Chr.1: 165544545-165544546 on Build GRCh38
rs138107763 Chr.1: 165544698-165544698 on Build GRCh38

View Full Product Details