Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109917074-109917347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF1
Entrez Gene ID
1435
Gene Name
colony stimulating factor 1
Gene Aliases
CSF-1, MCSF
UniGene
Hs.173894
Gene Chromosome Location
Chr.1: 109910506-109930994 on Build GRCh38
Transcripts
NM_000757.5 NM_172210.2 NM_172211.3 NM_172212.2 XM_017000369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4020448 c.253G>A p.A85T Chr.1: 109917320-109917320 on Build GRCh38
6191905 c.231T>G p.D77E Chr.1: 109917298-109917298 on Build GRCh38
6873561 c.279G>A p.M93I Chr.1: 109917346-109917346 on Build GRCh38
893587 c.262C>A p.L88M Chr.1: 109917329-109917329 on Build GRCh38
6873562 c.279G>A p.M93I Chr.1: 109917346-109917346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141065019 Chr.1: 109917310-109917310 on Build GRCh38
rs761742218 Chr.1: 109917311-109917311 on Build GRCh38
rs969747447 Chr.1: 109917152-109917152 on Build GRCh38
rs333967 Chr.1: 109917151-109917151 on Build GRCh38
rs763542778 Chr.1: 109917289-109917289 on Build GRCh38
rs534784288 Chr.1: 109917177-109917177 on Build GRCh38
rs994656171 Chr.1: 109917110-109917110 on Build GRCh38
rs776047822 Chr.1: 109917262-109917262 on Build GRCh38
rs1032787693 Chr.1: 109917180-109917180 on Build GRCh38
rs887371784 Chr.1: 109917242-109917242 on Build GRCh38
rs368995243 Chr.1: 109917138-109917138 on Build GRCh38
rs988967887 Chr.1: 109917199-109917199 on Build GRCh38
rs764578145 Chr.1: 109917304-109917304 on Build GRCh38
rs2297704 Chr.1: 109917220-109917220 on Build GRCh38
rs756814403 Chr.1: 109917115-109917115 on Build GRCh38
rs913830539 Chr.1: 109917119-109917119 on Build GRCh38
rs760439993 Chr.1: 109917256-109917256 on Build GRCh38
rs957369173 Chr.1: 109917187-109917187 on Build GRCh38
rs988045805 Chr.1: 109917111-109917111 on Build GRCh38
rs78075087 Chr.1: 109917200-109917200 on Build GRCh38
rs1001274049 Chr.1: 109917166-109917166 on Build GRCh38
rs370225963 Chr.1: 109917212-109917212 on Build GRCh38
rs932956712 Chr.1: 109917141-109917141 on Build GRCh38
rs375812519 Chr.1: 109917245-109917245 on Build GRCh38
rs3093044 Chr.1: 109917108-109917108 on Build GRCh38
rs915244178 Chr.1: 109917240-109917265 on Build GRCh38
rs1047322606 Chr.1: 109917241-109917241 on Build GRCh38
rs943168187 Chr.1: 109917193-109917193 on Build GRCh38
rs1013972451 Chr.1: 109917263-109917263 on Build GRCh38
rs987045233 Chr.1: 109917307-109917307 on Build GRCh38
rs770256756 Chr.1: 109917259-109917259 on Build GRCh38
rs745803514 Chr.1: 109917135-109917135 on Build GRCh38
rs1039877296 Chr.1: 109917214-109917214 on Build GRCh38
rs758791491 Chr.1: 109917283-109917283 on Build GRCh38
rs778506908 Chr.1: 109917126-109917126 on Build GRCh38

View Full Product Details