Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 47778685-47778924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKDC
Entrez Gene ID
5591
Gene Name
protein kinase, DNA-activated, catalytic polypeptide
Gene Aliases
DNA-PKcs, DNAPK, DNPK1, HYRC, HYRC1, IMD26, XRCC7, p350
UniGene
Hs.491682
Gene Chromosome Location
Chr.8: 47773108-47960183 on Build GRCh38
Transcripts
NM_001081640.1 NM_006904.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100171 c.11579G>A p.G3860D Chr.8: 47778797-47778797 on Build GRCh38
1644916 c.11616A>G p.K3872K Chr.8: 47778760-47778760 on Build GRCh38
1739458 c.11615A>G p.K3872R Chr.8: 47778761-47778761 on Build GRCh38
4717798 c.11604G>A p.T3868T Chr.8: 47778772-47778772 on Build GRCh38
1457247 c.11580C>T p.G3860G Chr.8: 47778796-47778796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8178246 Chr.8: 47778895-47778895 on Build GRCh38
rs780037710 Chr.8: 47778739-47778739 on Build GRCh38
rs878969227 Chr.8: 47778875-47778879 on Build GRCh38
rs550306617 Chr.8: 47778773-47778773 on Build GRCh38
rs749380423 Chr.8: 47778772-47778772 on Build GRCh38
rs568709506 Chr.8: 47778795-47778795 on Build GRCh38
rs377101643 Chr.8: 47778789-47778789 on Build GRCh38
rs764723005 Chr.8: 47778828-47778828 on Build GRCh38
rs754059871 Chr.8: 47778839-47778840 on Build GRCh38
rs941546183 Chr.8: 47778707-47778707 on Build GRCh38
rs193083438 Chr.8: 47778893-47778893 on Build GRCh38
rs761375915 Chr.8: 47778827-47778827 on Build GRCh38
rs774617314 Chr.8: 47778784-47778784 on Build GRCh38
rs538157705 Chr.8: 47778769-47778769 on Build GRCh38
rs554733258 Chr.8: 47778797-47778797 on Build GRCh38
rs192328175 Chr.8: 47778730-47778730 on Build GRCh38
rs869189873 Chr.8: 47778797-47778798 on Build GRCh38
rs750103974 Chr.8: 47778718-47778718 on Build GRCh38
rs975159802 Chr.8: 47778874-47778874 on Build GRCh38
rs746916693 Chr.8: 47778759-47778759 on Build GRCh38
rs908461708 Chr.8: 47778896-47778896 on Build GRCh38
rs1004525963 Chr.8: 47778849-47778864 on Build GRCh38
rs964142152 Chr.8: 47778864-47778864 on Build GRCh38
rs758213000 Chr.8: 47778720-47778720 on Build GRCh38
rs202104334 Chr.8: 47778796-47778796 on Build GRCh38
rs776045624 Chr.8: 47778709-47778709 on Build GRCh38
rs188253369 Chr.8: 47778880-47778880 on Build GRCh38
rs1017731965 Chr.8: 47778819-47778819 on Build GRCh38
rs373169292 Chr.8: 47778788-47778788 on Build GRCh38
rs183727221 Chr.8: 47778764-47778764 on Build GRCh38
rs374831588 Chr.8: 47778804-47778804 on Build GRCh38
rs753289499 Chr.8: 47778805-47778805 on Build GRCh38

View Full Product Details