Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2928843-2929080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLDA2
Entrez Gene ID
7262
Gene Name
pleckstrin homology like domain family A member 2
Gene Aliases
BRW1C, BWR1C, HLDA2, IPL, TSSC3
UniGene
Hs.154036
Gene Chromosome Location
Chr.11: 2928273-2929420 on Build GRCh38
Transcripts
NM_003311.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5512736 c.326A>T p.D109V Chr.11: 2929039-2929039 on Build GRCh38
6392789 c.404C>T p.S135F Chr.11: 2928961-2928961 on Build GRCh38
6384803 c.320T>C p.L107P Chr.11: 2929045-2929045 on Build GRCh38
7003800 c.349A>C p.T117P Chr.11: 2929016-2929016 on Build GRCh38
6425562 c.383C>T p.A128V Chr.11: 2928982-2928982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771457553 Chr.11: 2929002-2929002 on Build GRCh38
rs780442733 Chr.11: 2928890-2928890 on Build GRCh38
rs905284453 Chr.11: 2929040-2929040 on Build GRCh38
rs764476232 Chr.11: 2929016-2929016 on Build GRCh38
rs904539187 Chr.11: 2928997-2928997 on Build GRCh38
rs554127233 Chr.11: 2928932-2928932 on Build GRCh38
rs374438967 Chr.11: 2929028-2929028 on Build GRCh38
rs747774674 Chr.11: 2929055-2929055 on Build GRCh38
rs757832064 Chr.11: 2929008-2929008 on Build GRCh38
rs932101377 Chr.11: 2928909-2928909 on Build GRCh38
rs1001765868 Chr.11: 2929045-2929045 on Build GRCh38
rs756142635 Chr.11: 2928949-2928949 on Build GRCh38
rs890707017 Chr.11: 2928947-2928947 on Build GRCh38
rs759960240 Chr.11: 2929009-2929009 on Build GRCh38
rs763184811 Chr.11: 2928859-2928859 on Build GRCh38
rs752309318 Chr.11: 2929008-2929016 on Build GRCh38
rs945763216 Chr.11: 2928889-2928889 on Build GRCh38
rs199590203 Chr.11: 2929027-2929027 on Build GRCh38
rs35778007 Chr.11: 2928941-2928942 on Build GRCh38
rs778666265 Chr.11: 2928985-2928985 on Build GRCh38
rs770393406 Chr.11: 2929010-2929010 on Build GRCh38
rs371093380 Chr.11: 2928999-2928999 on Build GRCh38
rs200259411 Chr.11: 2929057-2929057 on Build GRCh38
rs753174881 Chr.11: 2928872-2928872 on Build GRCh38
rs762135434 Chr.11: 2929024-2929024 on Build GRCh38
rs749031914 Chr.11: 2928952-2928952 on Build GRCh38
rs912924956 Chr.11: 2928874-2928874 on Build GRCh38
rs757287852 Chr.11: 2928916-2928916 on Build GRCh38
rs917421059 Chr.11: 2928969-2928969 on Build GRCh38
rs778576348 Chr.11: 2928995-2929003 on Build GRCh38
rs754467448 Chr.11: 2928967-2928975 on Build GRCh38
rs778897407 Chr.11: 2929052-2929052 on Build GRCh38
rs756159463 Chr.11: 2929039-2929039 on Build GRCh38
rs149887098 Chr.11: 2929051-2929051 on Build GRCh38
rs775992906 Chr.11: 2929011-2929011 on Build GRCh38
rs558993627 Chr.11: 2928891-2928891 on Build GRCh38
rs751786648 Chr.11: 2928913-2928913 on Build GRCh38
rs768574034 Chr.11: 2928978-2928978 on Build GRCh38
rs751733546 Chr.11: 2929019-2929019 on Build GRCh38
rs765490981 Chr.11: 2928861-2928861 on Build GRCh38
rs753855394 Chr.11: 2929048-2929048 on Build GRCh38
rs939072844 Chr.11: 2928953-2928953 on Build GRCh38
rs772823181 Chr.11: 2929004-2929004 on Build GRCh38
rs764245340 Chr.11: 2928904-2928904 on Build GRCh38
rs779972526 Chr.11: 2929046-2929046 on Build GRCh38
rs777137816 Chr.11: 2929062-2929062 on Build GRCh38
rs376502838 Chr.11: 2928990-2928990 on Build GRCh38
rs944986204 Chr.11: 2928948-2928948 on Build GRCh38
rs745761657 Chr.11: 2928938-2928938 on Build GRCh38
rs536722787 Chr.11: 2929013-2929013 on Build GRCh38
rs779699187 Chr.11: 2928951-2928951 on Build GRCh38
rs771828334 Chr.11: 2929056-2929056 on Build GRCh38
rs572493288 Chr.11: 2928981-2928981 on Build GRCh38
rs925742769 Chr.11: 2928896-2928896 on Build GRCh38

View Full Product Details