Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13136003-13136250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NACC1
Entrez Gene ID
112939
Gene Name
nucleus accumbens associated 1
Gene Aliases
BEND8, BTBD14B, BTBD30, NAC-1, NAC1
UniGene
Hs.531614
Gene Chromosome Location
Chr.19: 13116848-13141147 on Build GRCh38
Transcripts
NM_052876.3 XM_005259721.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3528991 c.813C>T p.T271T Chr.19: 13136020-13136020 on Build GRCh38
2817268 c.873G>A p.E291E Chr.19: 13136080-13136080 on Build GRCh38
4513707 c.948C>T p.A316A Chr.19: 13136233-13136233 on Build GRCh38
7378827 c.868G>A p.E290K Chr.19: 13136075-13136075 on Build GRCh38
1216593 c.956T>C p.V319A Chr.19: 13136241-13136241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773852945 Chr.19: 13136173-13136173 on Build GRCh38
rs930814577 Chr.19: 13136181-13136181 on Build GRCh38
rs375417656 Chr.19: 13136193-13136196 on Build GRCh38
rs759463243 Chr.19: 13136194-13136194 on Build GRCh38
rs376033037 Chr.19: 13136211-13136211 on Build GRCh38
rs187770184 Chr.19: 13136163-13136163 on Build GRCh38
rs778988672 Chr.19: 13136140-13136140 on Build GRCh38
rs771547152 Chr.19: 13136232-13136232 on Build GRCh38
rs558296014 Chr.19: 13136144-13136144 on Build GRCh38
rs753751421 Chr.19: 13136112-13136112 on Build GRCh38
rs766198300 Chr.19: 13136100-13136100 on Build GRCh38
rs530200611 Chr.19: 13136047-13136047 on Build GRCh38
rs754906336 Chr.19: 13136119-13136119 on Build GRCh38
rs559883585 Chr.19: 13136038-13136038 on Build GRCh38
rs958808508 Chr.19: 13136116-13136116 on Build GRCh38
rs776238415 Chr.19: 13136056-13136056 on Build GRCh38
rs780416686 Chr.19: 13136219-13136219 on Build GRCh38
rs761461593 Chr.19: 13136074-13136074 on Build GRCh38
rs760172169 Chr.19: 13136053-13136053 on Build GRCh38
rs547805057 Chr.19: 13136020-13136020 on Build GRCh38
rs769523098 Chr.19: 13136193-13136193 on Build GRCh38
rs368552067 Chr.19: 13136181-13136181 on Build GRCh38
rs781035031 Chr.19: 13136188-13136188 on Build GRCh38
rs778137244 Chr.19: 13136152-13136152 on Build GRCh38
rs756559534 Chr.19: 13136218-13136218 on Build GRCh38
rs767089335 Chr.19: 13136080-13136080 on Build GRCh38
rs771675159 Chr.19: 13136178-13136178 on Build GRCh38
rs141390990 Chr.19: 13136029-13136029 on Build GRCh38
rs749840489 Chr.19: 13136222-13136222 on Build GRCh38
rs771401629 Chr.19: 13136158-13136158 on Build GRCh38
rs751784426 Chr.19: 13136209-13136209 on Build GRCh38
rs770206596 Chr.19: 13136170-13136170 on Build GRCh38
rs371748188 Chr.19: 13136202-13136202 on Build GRCh38
rs150402004 Chr.19: 13136143-13136143 on Build GRCh38
rs878950003 Chr.19: 13136023-13136023 on Build GRCh38
rs775502721 Chr.19: 13136218-13136219 on Build GRCh38
rs766124151 Chr.19: 13136189-13136189 on Build GRCh38
rs765076833 Chr.19: 13136196-13136196 on Build GRCh38
rs201155268 Chr.19: 13136086-13136086 on Build GRCh38
rs747276201 Chr.19: 13136156-13136156 on Build GRCh38
rs760367385 Chr.19: 13136188-13136188 on Build GRCh38
rs200464539 Chr.19: 13136191-13136191 on Build GRCh38
rs770541784 Chr.19: 13136176-13136199 on Build GRCh38
rs777031740 Chr.19: 13136024-13136024 on Build GRCh38
rs761210898 Chr.19: 13136177-13136177 on Build GRCh38
rs758379075 Chr.19: 13136200-13136200 on Build GRCh38
rs745650784 Chr.19: 13136192-13136199 on Build GRCh38
rs201327710 Chr.19: 13136233-13136233 on Build GRCh38
rs746717261 Chr.19: 13136170-13136170 on Build GRCh38
rs145116140 Chr.19: 13136164-13136164 on Build GRCh38
rs369677811 Chr.19: 13136212-13136212 on Build GRCh38
rs202215789 Chr.19: 13136199-13136199 on Build GRCh38
rs757582611 Chr.19: 13136210-13136210 on Build GRCh38
rs773764305 Chr.19: 13136058-13136058 on Build GRCh38
rs760455380 Chr.19: 13136092-13136092 on Build GRCh38

View Full Product Details