Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 146195069-146195336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLSCR4
Entrez Gene ID
57088
Gene Name
phospholipid scramblase 4
Gene Aliases
TRA1
UniGene
Hs.477869
Gene Chromosome Location
Chr.3: 146192335-146251179 on Build GRCh38
Transcripts
NM_001128304.1 NM_001128305.1 NM_001128306.1 NM_001177304.1 NM_020353.2 XM_005247654.2 XM_005247655.2 XM_011513031.2 XM_017006921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5697552 c.827T>C p.I276T Chr.3: 146195242-146195242 on Build GRCh38
3001951 c.818G>T p.G273V Chr.3: 146195251-146195251 on Build GRCh38
376800 c.831G>T p.R277R Chr.3: 146195238-146195238 on Build GRCh38
7148197 c.912G>A p.M304I Chr.3: 146195157-146195157 on Build GRCh38
4715848 c.817G>A p.G273S Chr.3: 146195252-146195252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147002895 Chr.3: 146195279-146195279 on Build GRCh38
rs140535240 Chr.3: 146195143-146195143 on Build GRCh38
rs764602258 Chr.3: 146195207-146195207 on Build GRCh38
rs769248215 Chr.3: 146195258-146195258 on Build GRCh38
rs746253409 Chr.3: 146195239-146195239 on Build GRCh38
rs139012801 Chr.3: 146195252-146195252 on Build GRCh38
rs767742285 Chr.3: 146195107-146195107 on Build GRCh38
rs143882752 Chr.3: 146195129-146195129 on Build GRCh38
rs768132771 Chr.3: 146195171-146195171 on Build GRCh38
rs751892195 Chr.3: 146195105-146195105 on Build GRCh38
rs150456352 Chr.3: 146195158-146195158 on Build GRCh38
rs146882263 Chr.3: 146195132-146195132 on Build GRCh38
rs778170967 Chr.3: 146195115-146195115 on Build GRCh38
rs759433696 Chr.3: 146195275-146195275 on Build GRCh38
rs543836526 Chr.3: 146195264-146195264 on Build GRCh38
rs753118891 Chr.3: 146195108-146195108 on Build GRCh38
rs16858016 Chr.3: 146195311-146195311 on Build GRCh38
rs79352490 Chr.3: 146195309-146195309 on Build GRCh38
rs189123579 Chr.3: 146195178-146195178 on Build GRCh38
rs767869749 Chr.3: 146195197-146195197 on Build GRCh38
rs966588083 Chr.3: 146195292-146195292 on Build GRCh38
rs759882317 Chr.3: 146195192-146195192 on Build GRCh38
rs764146798 Chr.3: 146195302-146195302 on Build GRCh38
rs377539047 Chr.3: 146195231-146195231 on Build GRCh38
rs763339276 Chr.3: 146195240-146195240 on Build GRCh38
rs755108318 Chr.3: 146195106-146195106 on Build GRCh38
rs752780039 Chr.3: 146195198-146195198 on Build GRCh38
rs149162101 Chr.3: 146195202-146195202 on Build GRCh38
rs770823235 Chr.3: 146195269-146195269 on Build GRCh38
rs201600148 Chr.3: 146195228-146195228 on Build GRCh38
rs959353383 Chr.3: 146195255-146195255 on Build GRCh38
rs779299265 Chr.3: 146195237-146195237 on Build GRCh38
rs747645523 Chr.3: 146195175-146195175 on Build GRCh38
rs34467190 Chr.3: 146195274-146195274 on Build GRCh38
rs143313993 Chr.3: 146195212-146195212 on Build GRCh38
rs753992152 Chr.3: 146195296-146195297 on Build GRCh38
rs779253488 Chr.3: 146195136-146195136 on Build GRCh38
rs138207609 Chr.3: 146195282-146195282 on Build GRCh38
rs762797844 Chr.3: 146195179-146195179 on Build GRCh38
rs375806529 Chr.3: 146195112-146195112 on Build GRCh38
rs138186262 Chr.3: 146195186-146195186 on Build GRCh38
rs370923591 Chr.3: 146195253-146195253 on Build GRCh38
rs748171162 Chr.3: 146195219-146195219 on Build GRCh38
rs764199832 Chr.3: 146195231-146195232 on Build GRCh38
rs879082910 Chr.3: 146195110-146195110 on Build GRCh38
rs774687691 Chr.3: 146195191-146195191 on Build GRCh38
rs1004984120 Chr.3: 146195096-146195096 on Build GRCh38
rs761013967 Chr.3: 146195107-146195107 on Build GRCh38
rs749598382 Chr.3: 146195127-146195127 on Build GRCh38
rs775587106 Chr.3: 146195280-146195280 on Build GRCh38
rs773200455 Chr.3: 146195259-146195259 on Build GRCh38
rs978432354 Chr.3: 146195304-146195304 on Build GRCh38

View Full Product Details