Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104766242-104766499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIPSNAP3B
Entrez Gene ID
55335
Gene Name
nipsnap homolog 3B
Gene Aliases
FP944, NIPSNAP3, SNAP1
UniGene
Hs.429294
Gene Chromosome Location
Chr.9: 104763741-104777764 on Build GRCh38
Transcripts
NM_018376.3 NR_130759.1 NR_130760.1 XM_011518839.1 XM_017014889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
298557 c.119G>A p.R40H Chr.9: 104766383-104766383 on Build GRCh38
4976492 c.96C>T p.Y32Y Chr.9: 104766360-104766360 on Build GRCh38
6479587 c.128A>G p.Y43C Chr.9: 104766392-104766392 on Build GRCh38
1459095 c.152C>T p.A51V Chr.9: 104766416-104766416 on Build GRCh38
4953277 c.154T>C p.F52L Chr.9: 104766418-104766418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005876510 Chr.9: 104766450-104766450 on Build GRCh38
rs911666323 Chr.9: 104766291-104766291 on Build GRCh38
rs370387843 Chr.9: 104766423-104766423 on Build GRCh38
rs768202178 Chr.9: 104766344-104766344 on Build GRCh38
rs780036326 Chr.9: 104766321-104766321 on Build GRCh38
rs200574459 Chr.9: 104766342-104766342 on Build GRCh38
rs117167547 Chr.9: 104766369-104766369 on Build GRCh38
rs527249516 Chr.9: 104766346-104766346 on Build GRCh38
rs183514449 Chr.9: 104766345-104766345 on Build GRCh38
rs199946236 Chr.9: 104766361-104766361 on Build GRCh38
rs137916198 Chr.9: 104766445-104766445 on Build GRCh38
rs992928251 Chr.9: 104766288-104766288 on Build GRCh38
rs145340292 Chr.9: 104766382-104766382 on Build GRCh38
rs768724264 Chr.9: 104766455-104766455 on Build GRCh38
rs1047320868 Chr.9: 104766409-104766409 on Build GRCh38
rs745465773 Chr.9: 104766296-104766296 on Build GRCh38
rs141665560 Chr.9: 104766374-104766374 on Build GRCh38
rs913429574 Chr.9: 104766294-104766294 on Build GRCh38
rs541747217 Chr.9: 104766329-104766329 on Build GRCh38
rs145226608 Chr.9: 104766383-104766383 on Build GRCh38
rs762345917 Chr.9: 104766464-104766464 on Build GRCh38
rs754503233 Chr.9: 104766385-104766385 on Build GRCh38
rs779207625 Chr.9: 104766282-104766282 on Build GRCh38
rs758052126 Chr.9: 104766325-104766325 on Build GRCh38
rs772451361 Chr.9: 104766358-104766358 on Build GRCh38
rs757999249 Chr.9: 104766378-104766378 on Build GRCh38
rs557871008 Chr.9: 104766451-104766451 on Build GRCh38
rs1010767444 Chr.9: 104766458-104766458 on Build GRCh38
rs377620680 Chr.9: 104766340-104766340 on Build GRCh38
rs776581790 Chr.9: 104766456-104766456 on Build GRCh38
rs777583472 Chr.9: 104766422-104766422 on Build GRCh38
rs775906437 Chr.9: 104766360-104766360 on Build GRCh38
rs764150779 Chr.9: 104766364-104766364 on Build GRCh38
rs370572755 Chr.9: 104766348-104766348 on Build GRCh38
rs769064472 Chr.9: 104766371-104766371 on Build GRCh38
rs754356351 Chr.9: 104766368-104766368 on Build GRCh38
rs570520079 Chr.9: 104766353-104766353 on Build GRCh38
rs555138221 Chr.9: 104766414-104766414 on Build GRCh38
rs761037348 Chr.9: 104766452-104766452 on Build GRCh38
rs749115055 Chr.9: 104766433-104766433 on Build GRCh38
rs905122676 Chr.9: 104766461-104766461 on Build GRCh38
rs565850921 Chr.9: 104766416-104766416 on Build GRCh38
rs576126992 Chr.9: 104766465-104766465 on Build GRCh38
rs144152053 Chr.9: 104766399-104766399 on Build GRCh38

View Full Product Details