Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64568697-64568930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A11
Entrez Gene ID
55867
Gene Name
solute carrier family 22 member 11
Gene Aliases
OAT4, hOAT4
UniGene
Hs.220844
Gene Chromosome Location
Chr.11: 64555601-64572875 on Build GRCh38
Transcripts
NM_001307985.1 NM_018484.3 XM_011545167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
267433 c.1341C>A p.C447* Chr.11: 64568737-64568737 on Build GRCh38
6780174 c.1382G>A p.R461Q Chr.11: 64568778-64568778 on Build GRCh38
6014479 c.1365C>G p.L455L Chr.11: 64568761-64568761 on Build GRCh38
6780171 c.1381C>T p.R461W Chr.11: 64568777-64568777 on Build GRCh38
1704208 c.1315G>A p.G439R Chr.11: 64568711-64568711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770592873 Chr.11: 64568805-64568805 on Build GRCh38
rs760597361 Chr.11: 64568779-64568780 on Build GRCh38
rs370867373 Chr.11: 64568904-64568904 on Build GRCh38
rs756192197 Chr.11: 64568771-64568771 on Build GRCh38
rs746680401 Chr.11: 64568735-64568735 on Build GRCh38
rs879618954 Chr.11: 64568883-64568883 on Build GRCh38
rs151223525 Chr.11: 64568767-64568767 on Build GRCh38
rs534852506 Chr.11: 64568899-64568899 on Build GRCh38
rs748473118 Chr.11: 64568794-64568794 on Build GRCh38
rs776152077 Chr.11: 64568744-64568744 on Build GRCh38
rs769325490 Chr.11: 64568749-64568749 on Build GRCh38
rs929749477 Chr.11: 64568741-64568741 on Build GRCh38
rs767805547 Chr.11: 64568769-64568769 on Build GRCh38
rs550281284 Chr.11: 64568882-64568882 on Build GRCh38
rs552406842 Chr.11: 64568895-64568895 on Build GRCh38
rs775329669 Chr.11: 64568756-64568756 on Build GRCh38
rs890803818 Chr.11: 64568768-64568768 on Build GRCh38
rs79107937 Chr.11: 64568843-64568844 on Build GRCh38
rs115000584 Chr.11: 64568889-64568889 on Build GRCh38
rs762811010 Chr.11: 64568820-64568820 on Build GRCh38
rs563654199 Chr.11: 64568891-64568892 on Build GRCh38
rs758922801 Chr.11: 64568748-64568748 on Build GRCh38
rs768645566 Chr.11: 64568830-64568830 on Build GRCh38
rs762943343 Chr.11: 64568761-64568761 on Build GRCh38
rs773188103 Chr.11: 64568771-64568773 on Build GRCh38
rs1009534752 Chr.11: 64568844-64568844 on Build GRCh38
rs754364564 Chr.11: 64568778-64568778 on Build GRCh38
rs779239657 Chr.11: 64568732-64568732 on Build GRCh38
rs779256060 Chr.11: 64568789-64568789 on Build GRCh38
rs530117390 Chr.11: 64568861-64568861 on Build GRCh38
rs1005213694 Chr.11: 64568846-64568846 on Build GRCh38
rs888075696 Chr.11: 64568776-64568776 on Build GRCh38
rs775050626 Chr.11: 64568819-64568819 on Build GRCh38
rs77918613 Chr.11: 64568898-64568898 on Build GRCh38
rs780871558 Chr.11: 64568812-64568812 on Build GRCh38
rs142153064 Chr.11: 64568738-64568738 on Build GRCh38
rs1015293269 Chr.11: 64568847-64568847 on Build GRCh38
rs751340166 Chr.11: 64568766-64568766 on Build GRCh38
rs769254236 Chr.11: 64568818-64568818 on Build GRCh38
rs866453080 Chr.11: 64568832-64568832 on Build GRCh38
rs561341898 Chr.11: 64568835-64568835 on Build GRCh38
rs368522762 Chr.11: 64568770-64568770 on Build GRCh38
rs780181907 Chr.11: 64568777-64568777 on Build GRCh38
rs998117006 Chr.11: 64568859-64568859 on Build GRCh38
rs745502502 Chr.11: 64568814-64568814 on Build GRCh38
rs532584329 Chr.11: 64568892-64568892 on Build GRCh38
rs764037518 Chr.11: 64568765-64568765 on Build GRCh38
rs770506670 Chr.11: 64568737-64568737 on Build GRCh38

View Full Product Details