Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29979112-29979347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAOK2
Entrez Gene ID
9344
Gene Name
TAO kinase 2
Gene Aliases
MAP3K17, PSK, PSK1, PSK1-BETA, TAO1, TAO2
UniGene
Hs.291623
Gene Chromosome Location
Chr.16: 29973641-29992261 on Build GRCh38
Transcripts
NM_001252043.1 NM_004783.3 NM_016151.3 XM_011545982.1 XM_011545983.2 XM_011545984.1 XM_011545985.1 XM_011545986.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
969701 c.553A>G p.T185A Chr.16: 29979298-29979298 on Build GRCh38
3932321 c.505G>A p.D169N Chr.16: 29979250-29979250 on Build GRCh38
5684586 c.450-1G>T p.? Chr.16: 29979194-29979194 on Build GRCh38
5684587 c.450-1G>T p.? Chr.16: 29979194-29979194 on Build GRCh38
703339 c.451G>T p.D151Y Chr.16: 29979196-29979196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977894084 Chr.16: 29979244-29979244 on Build GRCh38
rs562749630 Chr.16: 29979165-29979165 on Build GRCh38
rs201648224 Chr.16: 29979264-29979264 on Build GRCh38
rs373143692 Chr.16: 29979164-29979164 on Build GRCh38
rs776236834 Chr.16: 29979275-29979275 on Build GRCh38
rs771990943 Chr.16: 29979191-29979191 on Build GRCh38
rs940804292 Chr.16: 29979322-29979322 on Build GRCh38
rs745715123 Chr.16: 29979188-29979188 on Build GRCh38
rs774552364 Chr.16: 29979297-29979297 on Build GRCh38
rs140117945 Chr.16: 29979284-29979284 on Build GRCh38
rs768089730 Chr.16: 29979268-29979268 on Build GRCh38
rs763722316 Chr.16: 29979313-29979314 on Build GRCh38
rs533257570 Chr.16: 29979325-29979325 on Build GRCh38
rs760047779 Chr.16: 29979315-29979315 on Build GRCh38
rs919570391 Chr.16: 29979239-29979239 on Build GRCh38
rs78602377 Chr.16: 29979190-29979190 on Build GRCh38
rs939417987 Chr.16: 29979269-29979269 on Build GRCh38
rs778550779 Chr.16: 29979176-29979176 on Build GRCh38
rs746029734 Chr.16: 29979166-29979166 on Build GRCh38
rs570443018 Chr.16: 29979195-29979195 on Build GRCh38
rs756331599 Chr.16: 29979180-29979180 on Build GRCh38
rs756322108 Chr.16: 29979161-29979161 on Build GRCh38
rs746905198 Chr.16: 29979309-29979312 on Build GRCh38
rs778002938 Chr.16: 29979163-29979163 on Build GRCh38
rs369355810 Chr.16: 29979164-29979165 on Build GRCh38
rs764993691 Chr.16: 29979291-29979291 on Build GRCh38
rs752822328 Chr.16: 29979148-29979148 on Build GRCh38

View Full Product Details