Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33153476-33153707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUSD5
Entrez Gene ID
26032
Gene Name
sushi domain containing 5
Gene Aliases
-
UniGene
Hs.196647
Gene Chromosome Location
Chr.3: 33150041-33219215 on Build GRCh38
Transcripts
NM_015551.1 XM_005265034.3 XM_017006137.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4802276 c.1138G>A p.A380T Chr.3: 33153494-33153494 on Build GRCh38
5987890 c.956C>T p.S319F Chr.3: 33153676-33153676 on Build GRCh38
3357702 c.1041G>A p.S347S Chr.3: 33153591-33153591 on Build GRCh38
4116954 c.993A>C p.P331P Chr.3: 33153639-33153639 on Build GRCh38
730307 c.1058A>T p.K353M Chr.3: 33153574-33153574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75341012 Chr.3: 33153660-33153660 on Build GRCh38
rs764556693 Chr.3: 33153569-33153569 on Build GRCh38
rs781136711 Chr.3: 33153602-33153602 on Build GRCh38
rs193106795 Chr.3: 33153591-33153591 on Build GRCh38
rs901042723 Chr.3: 33153599-33153599 on Build GRCh38
rs765605768 Chr.3: 33153588-33153588 on Build GRCh38
rs770641633 Chr.3: 33153499-33153499 on Build GRCh38
rs575327243 Chr.3: 33153573-33153573 on Build GRCh38
rs773824741 Chr.3: 33153512-33153512 on Build GRCh38
rs1042894582 Chr.3: 33153655-33153655 on Build GRCh38
rs747145544 Chr.3: 33153631-33153631 on Build GRCh38
rs113725248 Chr.3: 33153592-33153592 on Build GRCh38
rs778706955 Chr.3: 33153607-33153607 on Build GRCh38
rs368207039 Chr.3: 33153632-33153632 on Build GRCh38
rs6810039 Chr.3: 33153498-33153498 on Build GRCh38
rs757019323 Chr.3: 33153574-33153575 on Build GRCh38
rs761040522 Chr.3: 33153562-33153562 on Build GRCh38
rs774020468 Chr.3: 33153633-33153633 on Build GRCh38
rs753164011 Chr.3: 33153550-33153550 on Build GRCh38
rs374212874 Chr.3: 33153518-33153518 on Build GRCh38
rs148017080 Chr.3: 33153647-33153647 on Build GRCh38
rs745331449 Chr.3: 33153644-33153644 on Build GRCh38
rs755874748 Chr.3: 33153611-33153611 on Build GRCh38
rs772120033 Chr.3: 33153521-33153521 on Build GRCh38
rs368497087 Chr.3: 33153551-33153551 on Build GRCh38
rs376105657 Chr.3: 33153612-33153612 on Build GRCh38
rs753483594 Chr.3: 33153564-33153564 on Build GRCh38
rs749142647 Chr.3: 33153618-33153618 on Build GRCh38
rs934567553 Chr.3: 33153676-33153676 on Build GRCh38
rs775997469 Chr.3: 33153657-33153657 on Build GRCh38
rs768025399 Chr.3: 33153542-33153542 on Build GRCh38
rs754320345 Chr.3: 33153572-33153572 on Build GRCh38
rs748050637 Chr.3: 33153603-33153603 on Build GRCh38

View Full Product Details