Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45723709-45723966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPINT4
Entrez Gene ID
391253
Gene Name
serine peptidase inhibitor, Kunitz type 4
Gene Aliases
C20orf137, SPINT3, dJ601O1.1
UniGene
Hs.677891
Gene Chromosome Location
Chr.20: 45722349-45725696 on Build GRCh38
Transcripts
NM_178455.1 XM_017027668.1

Gene Symbol
WFDC3
Entrez Gene ID
140686
Gene Name
WAP four-disulfide core domain 3
Gene Aliases
WAP14, dJ447F3.3
UniGene
Hs.419126
Gene Chromosome Location
Chr.20: 45708860-45791908 on Build GRCh38
Transcripts
NM_178455.1 XM_017027668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6856996 c.150C>A p.C50* Chr.20: 45723914-45723914 on Build GRCh38
1227388 c.164T>G p.F55C Chr.20: 45723928-45723928 on Build GRCh38
5984530 c.182G>C p.R61T Chr.20: 45723946-45723946 on Build GRCh38
5914329 c.188C>T p.S63F Chr.20: 45723952-45723952 on Build GRCh38
4601942 c.171T>A p.Y57* Chr.20: 45723935-45723935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748340638 Chr.20: 45723887-45723887 on Build GRCh38
rs375759176 Chr.20: 45723868-45723868 on Build GRCh38
rs192699400 Chr.20: 45723880-45723880 on Build GRCh38
rs1012778913 Chr.20: 45723745-45723745 on Build GRCh38
rs148466992 Chr.20: 45723874-45723874 on Build GRCh38
rs576441681 Chr.20: 45723799-45723799 on Build GRCh38
rs749305801 Chr.20: 45723906-45723906 on Build GRCh38
rs372041552 Chr.20: 45723861-45723861 on Build GRCh38
rs954586926 Chr.20: 45723820-45723820 on Build GRCh38
rs146246677 Chr.20: 45723784-45723784 on Build GRCh38
rs900179463 Chr.20: 45723795-45723795 on Build GRCh38
rs878897742 Chr.20: 45723929-45723929 on Build GRCh38
rs778588869 Chr.20: 45723862-45723862 on Build GRCh38
rs1016176223 Chr.20: 45723759-45723759 on Build GRCh38
rs763877419 Chr.20: 45723853-45723853 on Build GRCh38
rs1025995283 Chr.20: 45723792-45723792 on Build GRCh38
rs962999785 Chr.20: 45723768-45723768 on Build GRCh38
rs759988065 Chr.20: 45723932-45723932 on Build GRCh38
rs534751151 Chr.20: 45723845-45723845 on Build GRCh38
rs769960383 Chr.20: 45723894-45723894 on Build GRCh38
rs758416844 Chr.20: 45723869-45723869 on Build GRCh38
rs955811249 Chr.20: 45723823-45723823 on Build GRCh38
rs545308817 Chr.20: 45723882-45723882 on Build GRCh38
rs751965060 Chr.20: 45723843-45723843 on Build GRCh38
rs988632900 Chr.20: 45723836-45723836 on Build GRCh38
rs753405152 Chr.20: 45723857-45723857 on Build GRCh38
rs766801704 Chr.20: 45723829-45723829 on Build GRCh38
rs370520031 Chr.20: 45723895-45723895 on Build GRCh38
rs771483698 Chr.20: 45723907-45723907 on Build GRCh38
rs758887764 Chr.20: 45723746-45723746 on Build GRCh38
rs774718188 Chr.20: 45723923-45723923 on Build GRCh38
rs781524687 Chr.20: 45723883-45723883 on Build GRCh38

View Full Product Details