Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 20001184-20001437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PP2D1
Entrez Gene ID
151649
Gene Name
protein phosphatase 2C like domain containing 1
Gene Aliases
C3orf48
UniGene
Hs.733161
Gene Chromosome Location
Chr.3: 19979961-20012273 on Build GRCh38
Transcripts
NM_001252657.1 NR_027694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2948411 c.752T>C p.F251S Chr.3: 20001368-20001368 on Build GRCh38
1177218 c.769G>T p.E257* Chr.3: 20001351-20001351 on Build GRCh38
6215030 c.793C>T p.L265F Chr.3: 20001327-20001327 on Build GRCh38
7280628 c.923A>G p.Q308R Chr.3: 20001197-20001197 on Build GRCh38
4437594 c.706C>A p.P236T Chr.3: 20001414-20001414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010817348 Chr.3: 20001295-20001295 on Build GRCh38
rs770616864 Chr.3: 20001242-20001242 on Build GRCh38
rs117216652 Chr.3: 20001369-20001369 on Build GRCh38
rs978808773 Chr.3: 20001257-20001257 on Build GRCh38
rs4103004 Chr.3: 20001342-20001342 on Build GRCh38
rs184958519 Chr.3: 20001390-20001390 on Build GRCh38
rs866318489 Chr.3: 20001328-20001328 on Build GRCh38
rs755268420 Chr.3: 20001376-20001376 on Build GRCh38
rs965519841 Chr.3: 20001202-20001202 on Build GRCh38
rs531852426 Chr.3: 20001208-20001208 on Build GRCh38
rs867141007 Chr.3: 20001218-20001218 on Build GRCh38
rs149789258 Chr.3: 20001343-20001343 on Build GRCh38
rs182437145 Chr.3: 20001341-20001341 on Build GRCh38
rs967780707 Chr.3: 20001209-20001209 on Build GRCh38
rs765790338 Chr.3: 20001205-20001205 on Build GRCh38
rs893753974 Chr.3: 20001282-20001282 on Build GRCh38
rs533784654 Chr.3: 20001358-20001358 on Build GRCh38
rs766720647 Chr.3: 20001357-20001358 on Build GRCh38
rs199921992 Chr.3: 20001386-20001386 on Build GRCh38
rs199957786 Chr.3: 20001314-20001314 on Build GRCh38

View Full Product Details