Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231732855-231733128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE6D
Entrez Gene ID
5147
Gene Name
phosphodiesterase 6D
Gene Aliases
JBTS22, PDED
UniGene
Hs.516808
Gene Chromosome Location
Chr.2: 231732425-231781327 on Build GRCh38
Transcripts
NM_001291018.1 NM_002601.3 XM_011511342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1017776 c.380T>C p.V127A Chr.2: 231733025-231733025 on Build GRCh38
6039456 c.402delT p.F134fs*7 Chr.2: 231733003-231733003 on Build GRCh38
4092352 c.385A>G p.I129V Chr.2: 231733020-231733020 on Build GRCh38
5666580 c.372-2A>T p.? Chr.2: 231733035-231733035 on Build GRCh38
3048997 c.379G>A p.V127I Chr.2: 231733026-231733026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897600767 Chr.2: 231732927-231732927 on Build GRCh38
rs200295638 Chr.2: 231732958-231732958 on Build GRCh38
rs746367047 Chr.2: 231733069-231733069 on Build GRCh38
rs751628815 Chr.2: 231733043-231733043 on Build GRCh38
rs768190057 Chr.2: 231732955-231732955 on Build GRCh38
rs746056974 Chr.2: 231733090-231733090 on Build GRCh38
rs1010745157 Chr.2: 231733071-231733071 on Build GRCh38
rs753006146 Chr.2: 231733026-231733026 on Build GRCh38
rs778020664 Chr.2: 231733041-231733041 on Build GRCh38
rs758653268 Chr.2: 231733027-231733027 on Build GRCh38
rs201184663 Chr.2: 231732935-231732935 on Build GRCh38
rs780360512 Chr.2: 231733073-231733073 on Build GRCh38
rs1050511310 Chr.2: 231732913-231732913 on Build GRCh38
rs749335524 Chr.2: 231733069-231733070 on Build GRCh38
rs746149981 Chr.2: 231732919-231732919 on Build GRCh38
rs142298615 Chr.2: 231732999-231732999 on Build GRCh38
rs781613252 Chr.2: 231733067-231733067 on Build GRCh38
rs775174959 Chr.2: 231732936-231732936 on Build GRCh38
rs1005943411 Chr.2: 231732892-231732893 on Build GRCh38
rs770187087 Chr.2: 231732922-231732922 on Build GRCh38
rs781626399 Chr.2: 231732918-231732918 on Build GRCh38
rs146510084 Chr.2: 231732996-231732996 on Build GRCh38
rs755330705 Chr.2: 231733048-231733048 on Build GRCh38
rs35244888 Chr.2: 231732943-231732944 on Build GRCh38
rs770098446 Chr.2: 231733070-231733070 on Build GRCh38
rs755012805 Chr.2: 231732913-231732913 on Build GRCh38
rs373459369 Chr.2: 231733000-231733000 on Build GRCh38
rs376732436 Chr.2: 231733076-231733076 on Build GRCh38
rs761753448 Chr.2: 231732990-231732990 on Build GRCh38
rs747951123 Chr.2: 231733077-231733077 on Build GRCh38
rs547505745 Chr.2: 231732997-231732997 on Build GRCh38
rs199614325 Chr.2: 231732894-231732895 on Build GRCh38
rs777705791 Chr.2: 231733010-231733011 on Build GRCh38
rs978815158 Chr.2: 231733012-231733012 on Build GRCh38
rs757783742 Chr.2: 231733065-231733065 on Build GRCh38
rs762565226 Chr.2: 231732942-231732942 on Build GRCh38

View Full Product Details