Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2509926-2510199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PANK4
Entrez Gene ID
55229
Gene Name
pantothenate kinase 4
Gene Aliases
-
UniGene
Hs.26156
Gene Chromosome Location
Chr.1: 2508531-2526628 on Build GRCh38
Transcripts
NM_018216.2 XM_017001633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1340835 c.2033T>C p.V678A Chr.1: 2510063-2510063 on Build GRCh38
1241542 c.2002G>A p.V668M Chr.1: 2510094-2510094 on Build GRCh38
7091061 c.2012G>T p.R671L Chr.1: 2510084-2510084 on Build GRCh38
7348798 c.2012G>A p.R671H Chr.1: 2510084-2510084 on Build GRCh38
1260853 c.1950G>A p.A650A Chr.1: 2510146-2510146 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747087610 Chr.1: 2510048-2510048 on Build GRCh38
rs757217185 Chr.1: 2510129-2510129 on Build GRCh38
rs769191715 Chr.1: 2510141-2510141 on Build GRCh38
rs547687988 Chr.1: 2510119-2510119 on Build GRCh38
rs149944330 Chr.1: 2510077-2510077 on Build GRCh38
rs776726313 Chr.1: 2510054-2510055 on Build GRCh38
rs760823675 Chr.1: 2510028-2510030 on Build GRCh38
rs371537463 Chr.1: 2510090-2510090 on Build GRCh38
rs200709447 Chr.1: 2509946-2509946 on Build GRCh38
rs763642760 Chr.1: 2509955-2509955 on Build GRCh38
rs772790449 Chr.1: 2510174-2510174 on Build GRCh38
rs375754008 Chr.1: 2509971-2509971 on Build GRCh38
rs1036255012 Chr.1: 2510162-2510164 on Build GRCh38
rs774707895 Chr.1: 2510146-2510146 on Build GRCh38
rs143369621 Chr.1: 2510094-2510094 on Build GRCh38
rs897634191 Chr.1: 2510171-2510171 on Build GRCh38
rs755716268 Chr.1: 2510118-2510118 on Build GRCh38
rs59543907 Chr.1: 2510062-2510062 on Build GRCh38
rs751699001 Chr.1: 2510066-2510066 on Build GRCh38
rs752153802 Chr.1: 2510076-2510076 on Build GRCh38
rs767111444 Chr.1: 2510084-2510084 on Build GRCh38
rs2494619 Chr.1: 2509953-2509953 on Build GRCh38
rs578139474 Chr.1: 2509962-2509962 on Build GRCh38
rs780993278 Chr.1: 2510130-2510130 on Build GRCh38
rs762448493 Chr.1: 2510152-2510152 on Build GRCh38
rs749154729 Chr.1: 2510039-2510039 on Build GRCh38
rs779889996 Chr.1: 2510034-2510034 on Build GRCh38
rs752948285 Chr.1: 2509982-2509982 on Build GRCh38
rs561244718 Chr.1: 2510044-2510044 on Build GRCh38
rs563780919 Chr.1: 2510067-2510067 on Build GRCh38
rs375851673 Chr.1: 2510109-2510109 on Build GRCh38
rs777778664 Chr.1: 2510045-2510045 on Build GRCh38
rs754867219 Chr.1: 2510107-2510107 on Build GRCh38
rs778787395 Chr.1: 2510108-2510108 on Build GRCh38
rs771078250 Chr.1: 2510051-2510051 on Build GRCh38
rs201665819 Chr.1: 2510028-2510028 on Build GRCh38
rs527571212 Chr.1: 2510113-2510113 on Build GRCh38
rs753653285 Chr.1: 2510095-2510095 on Build GRCh38
rs373632754 Chr.1: 2510036-2510036 on Build GRCh38
rs570527995 Chr.1: 2510164-2510164 on Build GRCh38
rs1019063883 Chr.1: 2509987-2509987 on Build GRCh38
rs757283195 Chr.1: 2510112-2510112 on Build GRCh38
rs766466884 Chr.1: 2510027-2510028 on Build GRCh38
rs375430738 Chr.1: 2510061-2510061 on Build GRCh38
rs763508124 Chr.1: 2510072-2510072 on Build GRCh38
rs772382655 Chr.1: 2509953-2509953 on Build GRCh38
rs764730514 Chr.1: 2510075-2510075 on Build GRCh38
rs751137955 Chr.1: 2509958-2509958 on Build GRCh38
rs199728795 Chr.1: 2510018-2510018 on Build GRCh38
rs975838128 Chr.1: 2510078-2510078 on Build GRCh38
rs201306088 Chr.1: 2510085-2510085 on Build GRCh38
rs763561364 Chr.1: 2509954-2509954 on Build GRCh38
rs984697057 Chr.1: 2510028-2510028 on Build GRCh38
rs755847602 Chr.1: 2510025-2510025 on Build GRCh38

View Full Product Details