Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200881444-200881671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIL3
Entrez Gene ID
53938
Gene Name
peptidylprolyl isomerase like 3
Gene Aliases
CYPJ
UniGene
Hs.121076
Gene Chromosome Location
Chr.2: 200870907-200889333 on Build GRCh38
Transcripts
NM_032472.3 NM_130906.2 XM_005246651.3 XM_005246652.4 XM_011511358.2 XM_017004352.1 XM_017004353.1 XM_017004354.1 XM_017004355.1 XM_017004356.1 XM_017004357.1 XM_017004358.1 XM_017004359.1 XM_017004360.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5994369 c.173-9G>A p.? Chr.2: 200881497-200881497 on Build GRCh38
6510459 c.197G>T p.G66V Chr.2: 200881476-200881476 on Build GRCh38
6510460 c.185G>T p.G62V Chr.2: 200881476-200881476 on Build GRCh38
5994368 c.185-9G>A p.? Chr.2: 200881497-200881497 on Build GRCh38
1404431 c.207G>T p.K69N Chr.2: 200881454-200881454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935742065 Chr.2: 200881520-200881523 on Build GRCh38
rs773964710 Chr.2: 200881537-200881537 on Build GRCh38
rs189749739 Chr.2: 200881542-200881542 on Build GRCh38
rs1040241145 Chr.2: 200881557-200881557 on Build GRCh38
rs372032358 Chr.2: 200881478-200881478 on Build GRCh38
rs749147032 Chr.2: 200881504-200881504 on Build GRCh38
rs958937529 Chr.2: 200881560-200881560 on Build GRCh38
rs764035340 Chr.2: 200881490-200881494 on Build GRCh38
rs1015520384 Chr.2: 200881469-200881469 on Build GRCh38
rs977473853 Chr.2: 200881479-200881479 on Build GRCh38
rs561032777 Chr.2: 200881567-200881567 on Build GRCh38
rs768665366 Chr.2: 200881519-200881519 on Build GRCh38
rs985935757 Chr.2: 200881527-200881527 on Build GRCh38
rs528716993 Chr.2: 200881584-200881584 on Build GRCh38
rs557900557 Chr.2: 200881545-200881545 on Build GRCh38
rs763504334 Chr.2: 200881577-200881577 on Build GRCh38
rs917236322 Chr.2: 200881628-200881628 on Build GRCh38
rs1027828055 Chr.2: 200881553-200881553 on Build GRCh38
rs148778524 Chr.2: 200881482-200881482 on Build GRCh38
rs991487660 Chr.2: 200881611-200881611 on Build GRCh38
rs753818475 Chr.2: 200881515-200881516 on Build GRCh38
rs930019067 Chr.2: 200881610-200881610 on Build GRCh38

View Full Product Details