Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 183569027-183569294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCF2
Entrez Gene ID
4688
Gene Name
neutrophil cytosolic factor 2
Gene Aliases
NCF-2, NOXA2, P67-PHOX, P67PHOX
UniGene
Hs.587558
Gene Chromosome Location
Chr.1: 183555562-183590921 on Build GRCh38
Transcripts
NM_000433.3 NM_001127651.2 NM_001190789.1 NM_001190794.1 XM_005245207.1 XM_011509580.1 XM_011509581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4025930 c.693G>A p.P231P Chr.1: 183569162-183569162 on Build GRCh38
124687 c.692C>G p.P231R Chr.1: 183569163-183569163 on Build GRCh38
900828 c.673G>A p.A225T Chr.1: 183569182-183569182 on Build GRCh38
900827 c.705G>T p.E235D Chr.1: 183569150-183569150 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775491711 Chr.1: 183569220-183569220 on Build GRCh38
rs780715462 Chr.1: 183569212-183569212 on Build GRCh38
rs746618335 Chr.1: 183569194-183569194 on Build GRCh38
rs770372216 Chr.1: 183569198-183569198 on Build GRCh38
rs777409864 Chr.1: 183569189-183569189 on Build GRCh38
rs374145879 Chr.1: 183569103-183569103 on Build GRCh38
rs762845110 Chr.1: 183569223-183569223 on Build GRCh38
rs569623376 Chr.1: 183569258-183569258 on Build GRCh38
rs533070652 Chr.1: 183569244-183569244 on Build GRCh38
rs765699858 Chr.1: 183569214-183569214 on Build GRCh38
rs565319719 Chr.1: 183569239-183569239 on Build GRCh38
rs530584547 Chr.1: 183569270-183569270 on Build GRCh38
rs754192116 Chr.1: 183569182-183569182 on Build GRCh38
rs150595188 Chr.1: 183569152-183569152 on Build GRCh38
rs933144227 Chr.1: 183569082-183569082 on Build GRCh38
rs776449371 Chr.1: 183569101-183569101 on Build GRCh38
rs759471094 Chr.1: 183569102-183569102 on Build GRCh38
rs35971401 Chr.1: 183569104-183569104 on Build GRCh38
rs758545086 Chr.1: 183569253-183569253 on Build GRCh38
rs767530948 Chr.1: 183569237-183569237 on Build GRCh38
rs191249861 Chr.1: 183569254-183569254 on Build GRCh38
rs780442146 Chr.1: 183569173-183569173 on Build GRCh38
rs1019029773 Chr.1: 183569066-183569066 on Build GRCh38
rs758117639 Chr.1: 183569183-183569183 on Build GRCh38
rs762052296 Chr.1: 183569228-183569228 on Build GRCh38
rs144451516 Chr.1: 183569163-183569163 on Build GRCh38
rs540247375 Chr.1: 183569238-183569238 on Build GRCh38
rs750922183 Chr.1: 183569164-183569164 on Build GRCh38
rs774457345 Chr.1: 183569227-183569227 on Build GRCh38
rs762630439 Chr.1: 183569185-183569187 on Build GRCh38
rs769832894 Chr.1: 183569217-183569217 on Build GRCh38
rs377089938 Chr.1: 183569213-183569213 on Build GRCh38
rs757295495 Chr.1: 183569153-183569153 on Build GRCh38
rs763317358 Chr.1: 183569141-183569141 on Build GRCh38
rs893464438 Chr.1: 183569179-183569179 on Build GRCh38
rs761489200 Chr.1: 183569091-183569091 on Build GRCh38
rs981618260 Chr.1: 183569263-183569263 on Build GRCh38
rs756610730 Chr.1: 183569165-183569165 on Build GRCh38
rs764405660 Chr.1: 183569148-183569148 on Build GRCh38

View Full Product Details