Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 122313168-122313439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRRF
Entrez Gene ID
92399
Gene Name
mitochondrial ribosome recycling factor
Gene Aliases
MRFF, MTRRF, RRF
UniGene
Hs.368011
Gene Chromosome Location
Chr.9: 122264603-122323464 on Build GRCh38
Transcripts
NM_001173512.1 NM_138777.3 NM_199177.2 XM_005252302.3 XM_011519183.2 XM_011519184.2 XM_011519185.2 XM_011519186.1 XM_011519187.2 XM_017015286.1 XM_017015287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6182443 c.582G>T p.V194V Chr.9: 122313257-122313257 on Build GRCh38
3653865 c.669G>A p.K223K Chr.9: 122313344-122313344 on Build GRCh38
6715536 c.590C>T p.A197V Chr.9: 122313265-122313265 on Build GRCh38
5472644 c.634C>T p.R212C Chr.9: 122313309-122313309 on Build GRCh38
71692 c.651C>G p.N217K Chr.9: 122313326-122313326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748870392 Chr.9: 122313271-122313272 on Build GRCh38
rs778743486 Chr.9: 122313222-122313222 on Build GRCh38
rs374965751 Chr.9: 122313249-122313249 on Build GRCh38
rs540980673 Chr.9: 122313401-122313401 on Build GRCh38
rs777848212 Chr.9: 122313205-122313205 on Build GRCh38
rs199893960 Chr.9: 122313324-122313324 on Build GRCh38
rs771748626 Chr.9: 122313236-122313236 on Build GRCh38
rs758069537 Chr.9: 122313329-122313329 on Build GRCh38
rs745660111 Chr.9: 122313327-122313327 on Build GRCh38
rs780827149 Chr.9: 122313336-122313336 on Build GRCh38
rs780300689 Chr.9: 122313384-122313384 on Build GRCh38
rs41313845 Chr.9: 122313310-122313310 on Build GRCh38
rs760104543 Chr.9: 122313356-122313356 on Build GRCh38
rs761854494 Chr.9: 122313397-122313397 on Build GRCh38
rs776311769 Chr.9: 122313252-122313252 on Build GRCh38
rs745687806 Chr.9: 122313234-122313234 on Build GRCh38
rs749160870 Chr.9: 122313213-122313213 on Build GRCh38
rs147364697 Chr.9: 122313350-122313350 on Build GRCh38
rs145168590 Chr.9: 122313367-122313367 on Build GRCh38
rs775061261 Chr.9: 122313239-122313239 on Build GRCh38
rs201638253 Chr.9: 122313320-122313320 on Build GRCh38
rs374590931 Chr.9: 122313341-122313341 on Build GRCh38
rs2297483 Chr.9: 122313321-122313321 on Build GRCh38
rs746671272 Chr.9: 122313243-122313243 on Build GRCh38
rs753964595 Chr.9: 122313309-122313309 on Build GRCh38
rs895200895 Chr.9: 122313240-122313240 on Build GRCh38
rs115742525 Chr.9: 122313204-122313204 on Build GRCh38
rs774748137 Chr.9: 122313280-122313280 on Build GRCh38
rs753907096 Chr.9: 122313390-122313390 on Build GRCh38
rs372861942 Chr.9: 122313400-122313400 on Build GRCh38
rs553071081 Chr.9: 122313285-122313285 on Build GRCh38
rs150659802 Chr.9: 122313301-122313301 on Build GRCh38
rs571238975 Chr.9: 122313306-122313322 on Build GRCh38
rs575491969 Chr.9: 122313332-122313332 on Build GRCh38
rs768445789 Chr.9: 122313334-122313334 on Build GRCh38
rs752820940 Chr.9: 122313202-122313202 on Build GRCh38
rs766845811 Chr.9: 122313265-122313265 on Build GRCh38
rs764107948 Chr.9: 122313306-122313306 on Build GRCh38
rs139445941 Chr.9: 122313353-122313353 on Build GRCh38
rs368331157 Chr.9: 122313300-122313300 on Build GRCh38
rs151009758 Chr.9: 122313305-122313305 on Build GRCh38
rs562208604 Chr.9: 122313263-122313263 on Build GRCh38
rs774385418 Chr.9: 122313318-122313319 on Build GRCh38
rs757204051 Chr.9: 122313217-122313217 on Build GRCh38
rs772261297 Chr.9: 122313366-122313366 on Build GRCh38
rs760860243 Chr.9: 122313376-122313376 on Build GRCh38
rs890100993 Chr.9: 122313325-122313325 on Build GRCh38

View Full Product Details