Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99380044-99380295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ3
Entrez Gene ID
51805
Gene Name
coenzyme Q3, methyltransferase
Gene Aliases
DHHBMT, DHHBMTASE, UG0215E05, bA9819.1
UniGene
Hs.713623
Gene Chromosome Location
Chr.6: 99369401-99394223 on Build GRCh38
Transcripts
NM_017421.3 XM_005267018.4 XM_006715500.3 XM_011535898.2 XM_011535899.1 XM_017010933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082788 c.370A>G p.R124G Chr.6: 99380205-99380205 on Build GRCh38
3630720 c.356C>T p.S119F Chr.6: 99380219-99380219 on Build GRCh38
4486549 c.306C>T p.A102A Chr.6: 99380269-99380269 on Build GRCh38
4746058 c.368T>G p.L123R Chr.6: 99380207-99380207 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750880593 Chr.6: 99380173-99380173 on Build GRCh38
rs771884404 Chr.6: 99380252-99380252 on Build GRCh38
rs763723644 Chr.6: 99380258-99380258 on Build GRCh38
rs200544814 Chr.6: 99380222-99380222 on Build GRCh38
rs202070978 Chr.6: 99380217-99380217 on Build GRCh38
rs201503654 Chr.6: 99380262-99380262 on Build GRCh38
rs779285077 Chr.6: 99380157-99380157 on Build GRCh38
rs1047220123 Chr.6: 99380184-99380184 on Build GRCh38
rs757841060 Chr.6: 99380155-99380155 on Build GRCh38
rs753325156 Chr.6: 99380148-99380148 on Build GRCh38
rs267601176 Chr.6: 99380241-99380241 on Build GRCh38
rs775236131 Chr.6: 99380255-99380255 on Build GRCh38
rs780520402 Chr.6: 99380185-99380185 on Build GRCh38
rs923447747 Chr.6: 99380166-99380166 on Build GRCh38
rs371967196 Chr.6: 99380196-99380196 on Build GRCh38
rs769845397 Chr.6: 99380229-99380229 on Build GRCh38
rs976225059 Chr.6: 99380113-99380113 on Build GRCh38
rs755226826 Chr.6: 99380202-99380202 on Build GRCh38
rs745768147 Chr.6: 99380236-99380236 on Build GRCh38
rs1039043410 Chr.6: 99380080-99380081 on Build GRCh38
rs183814578 Chr.6: 99380180-99380180 on Build GRCh38
rs188980651 Chr.6: 99380233-99380233 on Build GRCh38
rs547078973 Chr.6: 99380102-99380102 on Build GRCh38
rs770996339 Chr.6: 99380141-99380141 on Build GRCh38

View Full Product Details