Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17366916-17367189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCR3LG1
Entrez Gene ID
374383
Gene Name
natural killer cell cytotoxicity receptor 3 ligand 1
Gene Aliases
B7-H6, B7H6, DKFZp686O24166
UniGene
Hs.146274
Gene Chromosome Location
Chr.11: 17351762-17377321 on Build GRCh38
Transcripts
NM_001202439.2 XM_011520072.2 XM_011520073.2 XM_011520074.2 XM_011520075.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5898412 c.553C>T p.P185S Chr.11: 17367140-17367140 on Build GRCh38
1730278 c.478A>T p.K160* Chr.11: 17367065-17367065 on Build GRCh38
4854202 c.428C>T p.P143L Chr.11: 17367015-17367015 on Build GRCh38
5717567 c.426C>T p.S142S Chr.11: 17367013-17367013 on Build GRCh38
5628880 c.482A>G p.Y161C Chr.11: 17367069-17367069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191529965 Chr.11: 17367159-17367159 on Build GRCh38
rs752317460 Chr.11: 17367045-17367045 on Build GRCh38
rs776130983 Chr.11: 17367086-17367086 on Build GRCh38
rs751519134 Chr.11: 17367022-17367022 on Build GRCh38
rs1005115969 Chr.11: 17366950-17366950 on Build GRCh38
rs147500995 Chr.11: 17367128-17367128 on Build GRCh38
rs963587010 Chr.11: 17367004-17367004 on Build GRCh38
rs952465220 Chr.11: 17366945-17366945 on Build GRCh38
rs989348977 Chr.11: 17367147-17367147 on Build GRCh38
rs753504321 Chr.11: 17367150-17367150 on Build GRCh38
rs976952464 Chr.11: 17367093-17367093 on Build GRCh38
rs759332868 Chr.11: 17367036-17367036 on Build GRCh38
rs923682964 Chr.11: 17367138-17367138 on Build GRCh38
rs932561292 Chr.11: 17367096-17367096 on Build GRCh38
rs1016191079 Chr.11: 17366986-17366986 on Build GRCh38
rs995818758 Chr.11: 17367006-17367006 on Build GRCh38
rs956264160 Chr.11: 17367140-17367140 on Build GRCh38
rs921186800 Chr.11: 17367095-17367095 on Build GRCh38
rs534920979 Chr.11: 17367092-17367092 on Build GRCh38
rs767472971 Chr.11: 17367041-17367041 on Build GRCh38
rs986132319 Chr.11: 17367118-17367118 on Build GRCh38
rs766047065 Chr.11: 17367018-17367018 on Build GRCh38
rs142133579 Chr.11: 17366991-17366992 on Build GRCh38
rs886315105 Chr.11: 17366943-17366943 on Build GRCh38
rs375121665 Chr.11: 17366962-17366962 on Build GRCh38
rs577650254 Chr.11: 17367002-17367002 on Build GRCh38
rs1044657942 Chr.11: 17367168-17367168 on Build GRCh38

View Full Product Details