Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177508116-177508385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK3
Entrez Gene ID
79930
Gene Name
docking protein 3
Gene Aliases
DOKL
UniGene
Hs.720849
Gene Chromosome Location
Chr.5: 177501904-177510507 on Build GRCh38
Transcripts
NM_001144875.1 NM_001144876.1 NM_001308235.1 NM_001308236.1 NM_024872.2 XM_005265985.3 XM_005265987.1 XM_011534654.2 XM_011534655.2 XM_011534656.2 XM_011534657.2 XM_017009883.1 XM_017009884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6364611 c.511A>G p.M171V Chr.5: 177508266-177508266 on Build GRCh38
7280324 c.436C>T p.R146W Chr.5: 177508341-177508341 on Build GRCh38
7280325 c.268C>T p.R90W Chr.5: 177508341-177508341 on Build GRCh38
7365296 c.256G>T p.E86* Chr.5: 177508353-177508353 on Build GRCh38
7365295 c.424G>T p.E142* Chr.5: 177508353-177508353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752829173 Chr.5: 177508356-177508356 on Build GRCh38
rs375564754 Chr.5: 177508308-177508308 on Build GRCh38
rs778804038 Chr.5: 177508325-177508325 on Build GRCh38
rs760143673 Chr.5: 177508349-177508349 on Build GRCh38
rs766116722 Chr.5: 177508213-177508213 on Build GRCh38
rs778697790 Chr.5: 177508363-177508363 on Build GRCh38
rs754060367 Chr.5: 177508360-177508360 on Build GRCh38
rs556129531 Chr.5: 177508228-177508228 on Build GRCh38
rs1028870894 Chr.5: 177508184-177508184 on Build GRCh38
rs745849199 Chr.5: 177508329-177508329 on Build GRCh38
rs565362873 Chr.5: 177508345-177508345 on Build GRCh38
rs751304422 Chr.5: 177508223-177508223 on Build GRCh38
rs368466434 Chr.5: 177508238-177508238 on Build GRCh38
rs755077501 Chr.5: 177508292-177508292 on Build GRCh38
rs1038481942 Chr.5: 177508172-177508172 on Build GRCh38
rs898544474 Chr.5: 177508177-177508177 on Build GRCh38
rs111533164 Chr.5: 177508332-177508332 on Build GRCh38
rs755787560 Chr.5: 177508176-177508176 on Build GRCh38
rs775080614 Chr.5: 177508229-177508229 on Build GRCh38
rs964755829 Chr.5: 177508327-177508327 on Build GRCh38
rs76251313 Chr.5: 177508158-177508158 on Build GRCh38
rs752160007 Chr.5: 177508279-177508279 on Build GRCh38
rs752814202 Chr.5: 177508297-177508297 on Build GRCh38
rs762896466 Chr.5: 177508205-177508205 on Build GRCh38
rs398000034 Chr.5: 177508220-177508220 on Build GRCh38
rs376371500 Chr.5: 177508237-177508237 on Build GRCh38
rs749124352 Chr.5: 177508203-177508203 on Build GRCh38
rs949192537 Chr.5: 177508354-177508354 on Build GRCh38
rs148416579 Chr.5: 177508333-177508333 on Build GRCh38
rs767648183 Chr.5: 177508353-177508353 on Build GRCh38
rs566438113 Chr.5: 177508151-177508151 on Build GRCh38
rs949244539 Chr.5: 177508142-177508142 on Build GRCh38
rs369855241 Chr.5: 177508335-177508335 on Build GRCh38
rs766977751 Chr.5: 177508278-177508278 on Build GRCh38
rs887828416 Chr.5: 177508309-177508309 on Build GRCh38
rs774341441 Chr.5: 177508220-177508220 on Build GRCh38
rs201649391 Chr.5: 177508212-177508212 on Build GRCh38
rs759037133 Chr.5: 177508222-177508222 on Build GRCh38
rs55761704 Chr.5: 177508215-177508215 on Build GRCh38
rs754603868 Chr.5: 177508225-177508225 on Build GRCh38
rs1045770050 Chr.5: 177508145-177508145 on Build GRCh38
rs753928803 Chr.5: 177508334-177508335 on Build GRCh38
rs750801141 Chr.5: 177508274-177508274 on Build GRCh38
rs372201708 Chr.5: 177508305-177508305 on Build GRCh38
rs952079142 Chr.5: 177508148-177508148 on Build GRCh38
rs771518601 Chr.5: 177508331-177508331 on Build GRCh38
rs975990351 Chr.5: 177508175-177508175 on Build GRCh38
rs150922665 Chr.5: 177508341-177508341 on Build GRCh38
rs770908544 Chr.5: 177508210-177508210 on Build GRCh38
rs1018000249 Chr.5: 177508324-177508324 on Build GRCh38
rs372101796 Chr.5: 177508194-177508194 on Build GRCh38
rs756373791 Chr.5: 177508301-177508301 on Build GRCh38
rs147721747 Chr.5: 177508312-177508312 on Build GRCh38
rs910562816 Chr.5: 177508159-177508159 on Build GRCh38
rs540258218 Chr.5: 177508339-177508339 on Build GRCh38
rs997042673 Chr.5: 177508179-177508179 on Build GRCh38
rs774957822 Chr.5: 177508346-177508346 on Build GRCh38
rs1004214493 Chr.5: 177508167-177508167 on Build GRCh38
rs142573859 Chr.5: 177508340-177508340 on Build GRCh38
rs763293316 Chr.5: 177508277-177508277 on Build GRCh38
rs533788535 Chr.5: 177508154-177508154 on Build GRCh38
rs767738059 Chr.5: 177508296-177508296 on Build GRCh38
rs143751041 Chr.5: 177508189-177508189 on Build GRCh38
rs756218542 Chr.5: 177508357-177508357 on Build GRCh38
rs757018092 Chr.5: 177508362-177508362 on Build GRCh38
rs1002425691 Chr.5: 177508230-177508230 on Build GRCh38

View Full Product Details